Skip to Content
Merck
All Photos(2)

Key Documents

224480

Sigma-Aldrich

2-Nitrocyclohexanone

99%

Synonym(s):

α-Nitrocyclohexanone, 2-Nitrocyclohexan-1-one, 2-Nitrocyclohexanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H9(=O)
CAS Number:
Molecular Weight:
143.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

39-43 °C (lit.)

functional group

ketone
nitro

SMILES string

[O-][N+](=O)C1CCCCC1=O

InChI

1S/C6H9NO3/c8-6-4-2-1-3-5(6)7(9)10/h5H,1-4H2

InChI key

SZNILIWUUKKNPE-UHFFFAOYSA-N

General description

The tautomerization rates of 2-nitrocyclohexanone (2-NCH) has been studied spectrophotometrically in several organic aprotic solvents and their binary mixtures.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gabriella Siani et al.
Organic & biomolecular chemistry, 6(22), 4236-4241 (2008-10-31)
The rates of tautomerization of 2-nitrocyclohexanone (2-NCH) have been measured spectrophotometrically at 25.0 +/- 0.1 degrees C in several organic aprotic solvents and their binary mixtures. In cyclohexane the reaction is effectively catalyzed by bases and inhibited by acids while

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service