Skip to Content
Merck
All Photos(4)

Key Documents

223832

Sigma-Aldrich

Bis(triphenylphosphoranylidene)ammonium chloride

97%

Synonym(s):

Bis(triphenylphosphine)iminium chloride, Hexaphenyldiphosphazenium chloride, PPNCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3P=N(Cl)=P(C6H5)3
CAS Number:
Molecular Weight:
574.03
Beilstein:
4097979
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

270-272 °C (lit.)

SMILES string

ClP(N=P(c1ccccc1)(c2ccccc2)c3ccccc3)(c4ccccc4)(c5ccccc5)c6ccccc6

InChI

1S/C36H30ClNP2/c37-40(34-25-13-4-14-26-34,35-27-15-5-16-28-35,36-29-17-6-18-30-36)38-39(31-19-7-1-8-20-31,32-21-9-2-10-22-32)33-23-11-3-12-24-33/h1-30H

InChI key

QMTYWEYUVVWPQL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Co-catalyst used with a chiral Cobalt-salen complex for the asymmetric addition of carbon dioxide to propylene oxide.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edward D Burgoyne et al.
The Analyst, 145(21), 7000-7008 (2020-09-02)
Miniaturization of electrochemical detection methods for point-of-care-devices is ideal for their integration and use within healthcare environments. Simultaneously, the prolific pathogenic bacteria Pseudomonas aeruginosa poses a serious health risk to patients with compromised immune systems. Recognizing these two factors, a
Albrecht Berkessel et al.
Organic letters, 8(20), 4401-4404 (2006-09-22)
New chiral catalyst systems were developed for the reaction of carbon dioxide with propylene oxide (PO) at atmospheric pressure to generate enantiomerically enriched propylene carbonate (PC). The best selectivity was achieved with a Co(III)(salen)-trifluoroacetyl complex and bis(triphenylphosphoranylidene)ammonium fluoride (PPN+F-) as
R Aubeck et al.
The Analyst, 116(8), 811-814 (1991-08-01)
The properties of an ion-selective electrode for indomethacin based on the bis(triphenylphosphoranylidene)ammonium-indomethacin ion-pair complex in a poly(vinyl chloride) membrane are described. The detection limit found for indomethacin at pH 7.0 was 2 x 10(-5) mol l-1 (6.4 micrograms ml-1) and
Carine Robert et al.
Nature communications, 2, 586-586 (2011-12-14)
The vast majority of commodity materials are obtained from petrochemical feedstocks. These resources will plausibly be depleted within the next 100 years, and the peak in global oil production is estimated to occur within the next few decades. In this
Evgeny Smirnov et al.
ACS nano, 9(6), 6565-6575 (2015-06-04)
Soft or "liquid-liquid" interfaces were functionalized by roughly half a monolayer of mirror-like nanofilms of gold nanoparticles using a precise interfacial microinjection method. The surface coverage of the nanofilm was characterized by ion transfer voltammetry. These gold nanoparticle films represent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service