Skip to Content
Merck
All Photos(2)

Key Documents

222526

Sigma-Aldrich

3-Methylxanthine

98%

Synonym(s):

2,6-Dihydroxy-3-methylpurine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N4O2
CAS Number:
Molecular Weight:
166.14
Beilstein:
180944
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

>300 °C (lit.)

SMILES string

CN1C(=O)NC(=O)c2[nH]cnc12

InChI

1S/C6H6N4O2/c1-10-4-3(7-2-8-4)5(11)9-6(10)12/h2H,1H3,(H,7,8)(H,9,11,12)

InChI key

GMSNIKWWOQHZGF-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

3-Methylxanthine, a metabolite of theophylline, was quantitated in rat plasma by a sensitive LC-MS/MS method.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Sang Won Lee et al.
Biochemistry, 49(13), 2943-2951 (2010-03-11)
RNAs often exhibit a high degree of conformational dynamics and heterogeneity, leading to a rugged energy landscape. However, the roles of conformational heterogeneity and rapid dynamics in molecular recognition or RNA function have not been extensively elucidated. Ultrafast time-resolved fluorescence
Sandrine Derkenne et al.
Pharmacogenetics and genomics, 15(7), 503-511 (2005-06-23)
Pharmacokinetics of theophylline was investigated in Cyp1(+/+) wild-type mice, Cyp1a1(-/-) and Cyp1a2(-/-) knockout mice, and humanized hCYP1A1_1A2 mice lacking either the mouse Cyp1a1 or Cyp1a2 gene. Animals received a single dose of theophylline (8 mg/kg i.p.), either alone or pretreated
H Konishi et al.
European journal of clinical pharmacology, 46(4), 309-312 (1994-01-01)
The effect of fluconazole, an antimycotic that inhibits cytochrome P-450-mediated drug metabolism, on theophylline kinetics and the production of its metabolites were compared with those of enoxacin in 5 healthy subjects. All subjects received a single oral dose of 240
M Sarkar et al.
Antimicrobial agents and chemotherapy, 34(4), 594-599 (1990-04-01)
Some quinolone antibiotics cause increases in levels of theophylline in plasma that lead to serious adverse effects. We investigated the mechanism of this interaction by developing an in vitro system of human liver microsomes. Theophylline (1,3-dimethylxanthine) was incubated with human
K Ueno et al.
DICP : the annals of pharmacotherapy, 25(7-8), 727-730 (1991-07-01)
The mechanism of an interaction between theophylline and mexiletine hydrochloride was investigated in 6 male inpatients coadministered both drugs and 16 inpatients (13 men, 3 women) administered theophylline only. Serum theophylline and mexiletine concentrations and urinary concentrations of theophylline and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service