Skip to Content
Merck
All Photos(1)

Key Documents

22135

Sigma-Aldrich

(+)-Cedrol

≥99.0% (sum of enantiomers, GC)

Synonym(s):

(1S,2R,5S,7R,8R)-2,6,6,8-Tetramethyltricyclo[5.3.1.01.5]undecan-8-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H26O
CAS Number:
Molecular Weight:
222.37
Beilstein:
2206347
EC Number:
MDL number:
UNSPSC Code:
12352212
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (sum of enantiomers, GC)

form

solid

optical activity

[α]20/D +10.5±1°, c = 5% in chloroform

bp

273 °C (lit.)

mp

82-86 °C
86-87 °C (lit.)

functional group

hydroxyl

SMILES string

C[C@@H]1CC[C@H]2C(C)(C)[C@H]3C[C@@]12CC[C@@]3(C)O

InChI

1S/C15H26O/c1-10-5-6-11-13(2,3)12-9-15(10,11)8-7-14(12,4)16/h10-12,16H,5-9H2,1-4H3/t10-,11+,12-,14-,15+/m1/s1

InChI key

SVURIXNDRWRAFU-OGMFBOKVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(+)-Cedrol is a crystalline hydrated product of α-cedrene, which is a sesquiterpene found in cedar-wood oil. It can be used as a fragrance ingredient in cosmetics, shampoos, and soaps as well as in non-cosmetic products such as cleaners and detergents. (±) Cedrol can be synthesized by intramolecular Diels-Alder reaction of alkyl cyclopentadiene.

Application

(+)-Cedrol can be used as a starting material for the preparation of cedryl acetate by acetylation using acetic anhydride in the presence of an acid catalyst. It can also be incorporated as a precursor for the total synthesis of rare illicium sesquiterpene (+)-pseudoanisatin via selective C-H bond functionalization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oscar Mbare et al.
BMC infectious diseases, 19(1), 800-800 (2019-09-13)
Larviciding is an effective supplementary tool for malaria vector control, but the identification and accessibility of aquatic habitats impedes application. Dissemination of the insect growth regulator, pyriproxyfen (PPF), by gravid Anopheles might constitute a novel application strategy. This study aimed
Catalytic acetylation of (+)-cedrol with heterogeneous catalyst H2SO4/SiO2 under solvent free conditions
Elvia R, et al.
Chemical Intermediates, 1(4), 196-201 (2015)
Kevin Hung et al.
Journal of the American Chemical Society, 138(51), 16616-16619 (2016-12-15)
Illicium sesquiterpenes have been the subject of numerous synthetic efforts due to their ornate and highly oxidized structures as well as significant biological activities. Herein we report the first chemical synthesis of (+)-pseudoanisatin from the abundant feedstock chemical cedrol (∼$50
Total synthesis of (.+-.)-cedrol and (.+-.)-cedrene via an intramolecular Diels-Alder reaction
Breitholle EG and Fallis AG
The Journal of Organic Chemistry, 43(10), 1964-1968 (1978)
Mitsuo Miyazawa et al.
Natural product research, 17(5), 313-317 (2003-10-07)
Microbial transformation of (+)-cedrol was investigated by using Staphylococcus epidermidis and found that stereospecific hydroxylation of (+)-cedrol occurred at the C-3 position to form (+)-(3S)-3-hydroxycedrol.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service