Skip to Content
Merck
All Photos(3)

Key Documents

220574

Sigma-Aldrich

(R)-(+)-α-Methylbenzyl isocyanate

99%

Synonym(s):

(R)-(+)-1-Phenylethyl isocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH3)NCO
CAS Number:
Molecular Weight:
147.17
Beilstein:
3196930
EC Number:
MDL number:
UNSPSC Code:
12352118
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

optical activity

[α]20/D +10°, neat

optical purity

ee: 96% (GLC)

refractive index

n20/D 1.513 (lit.)

bp

55-56 °C/2.5 mmHg (lit.)

density

1.045 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

C[C@@H](N=C=O)c1ccccc1

InChI

1S/C9H9NO/c1-8(10-7-11)9-5-3-2-4-6-9/h2-6,8H,1H3/t8-/m1/s1

InChI key

JJSCUXAFAJEQGB-MRVPVSSYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(R)-(+)-α-Methylbenzyl isocyanate is an organic compound that belongs to the class of isocyanates. It is used in the synthesis of chiral ligands for asymmetric catalysis. These ligands play a crucial role in enantioselective reactions, allowing to produce single enantiomer products.

Application

  • Paper Title: Stereoselective metabolism of nicotine and tobacco-specific N-nitrosamines to 4-hydroxy-4-(3-pyridyl)butanoic acid in rats. (Trushin N, Hecht SS, 1999).
  • Paper Title: Absolute configuration of 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanol formed metabolically from 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone. (Hecht SS, Spratt TE, Trushin N, 1997).

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-performance liquid chromatographic determination of the enantiomers of beta-adrenoceptor blocking agents in biological fluids. II. Studies with atenolol.
S K Chin et al.
Journal of chromatography, 489(2), 438-445 (1989-04-14)
Myriam Matoga et al.
Journal of enzyme inhibition and medicinal chemistry, 17(6), 375-379 (2003-04-10)
The derivatization of racemic 5-[(2-methylphenoxy)methyl]-2-amino-2-oxazoline, developed as an imidazoline binding sites ligand, with (+)-(R)-alpha-methylbenzyl isocyanate was performed in chloroform. The reaction led to two pairs of diastereomers, which were separated by RP-HPLC. A kinetic study of the derivatization reaction was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service