217808
cis-Aconitic anhydride
95%
Synonym(s):
cis-Propene-1,2,3-tricarboxylic anhydride
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C6H4O5
CAS Number:
Molecular Weight:
156.09
Beilstein:
127575
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Quality Level
Assay
95%
mp
75-78 °C (lit.)
SMILES string
OC(=O)CC1=CC(=O)OC1=O
InChI
1S/C6H4O5/c7-4(8)1-3-2-5(9)11-6(3)10/h2H,1H2,(H,7,8)
InChI key
GVJRTUUUJYMTNQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Ming Zhao et al.
Natural product communications, 7(6), 785-787 (2012-07-24)
From the seeds of Alisma orientale, cis-aconitic anhydride ethyl ester and cis-2,4,5-trihydroxycinnamic acid were isolated, together with nine known phenolic compounds and a megastigmane sesquiterpene. All compounds are reported for the first time from Alisma species.
M Wirth et al.
Pharmaceutical research, 15(7), 1031-1037 (1998-08-04)
To investigate the usefulness of wheat germ agglutinin as a targeting carrier protein for an acid-labile chemotherapeutic prodrug directed against colon carcinoma cells in vitro. Cis-aconityl-linked doxorubicin-wheat germ agglutinin was prepared by a two step procedure and the conjugate-binding capacity
A V Martynov et al.
Journal of interferon & cytokine research : the official journal of the International Society for Interferon and Cytokine Research, 25(7), 414-417 (2005-07-19)
The antiproliferative activity of aconitylated (AIFN) and succinylated (SIFN) derivatives of recombinant interferon- alpha2b (IFN-alpha2b) was examined. Acylation of IFN-alpha2b was performed by succinic and cis-aconitic anhydrides. Antiproliferative properties of AIFN and SIFN were studied in vitro on the CaOv
Peng-Fei Cui et al.
Journal of controlled release : official journal of the Controlled Release Society, 303, 253-262 (2019-04-28)
Cationic polymeric vectors attracted plenty of attentions in gene therapy due to nonimmunogenicity, easy to synthesis and flexible properties. However, biocompatibility challenge such as nonspecific interactions with blood cells and serum proteins, may affect the delivery efficiency of cationic vectors;
Abhijeet Pandey et al.
Materials science & engineering. C, Materials for biological applications, 98, 419-436 (2019-03-01)
In the present investigation, FePt alloy nanoparticles were synthesized with controlled size and elemental composition followed by surface modification using (3-Aminopropyl) triethoxysilane (APTES). Lenalidomide was covalently bound to FePt-NH2 by pH sensitive hydrazone bonding. Hyaluronic acid was conjugated to amino
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service