Skip to Content
Merck
All Photos(2)

Key Documents

216836

Sigma-Aldrich

Tolylene-2,4-diisocyanate

technical grade, 80%

Synonym(s):

2,4-Diisocyanatotoluene, 4-Methyl-1,3-phenylene diisocyanate, 4-Methyl-m-phenylene diisocyanate, BASF LUPRANATE T80, TDI

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NCO)2
CAS Number:
Molecular Weight:
174.16
Beilstein:
744602
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

vapor density

6 (vs air)

vapor pressure

0.03 mmHg ( 25 °C)

Assay

80%

autoignition temp.

>1148 °F

expl. lim.

9.5 %

refractive index

n20/D 1.568 (lit.)

bp

115-120 °C/10 mmHg (lit.)

mp

20-22 °C (lit.)

solubility

acetone and ethanol: soluble(lit.)
alcohol: miscible(lit.)
benzene: soluble(lit.)
carbon tetrachloride: miscible(lit.)
chlorobenzene: miscible(lit.)
diethyl ether: miscible(lit.)
kerosene: miscible(lit.)
olive oil: miscible(lit.)

density

1.225 g/mL at 25 °C

functional group

isocyanate

storage temp.

2-8°C

SMILES string

Cc1ccc(cc1N=C=O)N=C=O

InChI

1S/C9H6N2O2/c1-7-2-3-8(10-5-12)4-9(7)11-6-13/h2-4H,1H3

InChI key

DVKJHBMWWAPEIU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Remainder tolylene 2,6-diisocyanate

Legal Information

Product of BASF AG (Aktiengesellschaft)

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

269.6 °F - closed cup

Flash Point(C)

132 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhenbin Zhang et al.
Methods in molecular biology (Clifton, N.J.), 970, 457-467 (2013-01-04)
Capillary electrochromatography (CEC) has been shown to be a promising technique for miniaturized enantioseparations. Here we present the preparation and application of positively charged cellulose derivative-based chiral stationary phases (CSPs) for the CEC separation of enantiomers. The cellulose derivatives were
Gabriel Kiss et al.
Polymers, 12(7) (2020-07-16)
Ester polyurethane (PU) foam waste was reacted at atmospheric pressure in an autoclave and using microwaves with diethylene glycol (DEG) at different PU/DEG ratios in the presence of diethanolamine as a catalyst to find the glycolysis conditions that allow for
Zhifang Yang et al.
Langmuir : the ACS journal of surfaces and colloids, 26(12), 9386-9392 (2010-04-29)
A novel ABA triblock copolymer containing electroactive tetraaniline [(ANI)(4)] and poly(ethylene oxide) (PEO600, M(n) = 600), (ANI)(4)-b-PEO600-b-(ANI)(4), was synthesized by coupling tetraaniline and PEO600 with tolylene 2,4-diisocyanate. FTIR, NMR, and UV-vis spectroscopy were combined to characterize the chemical structure of
Effects of solvent polarity on the reaction of phenol with tolylene-2, 4-diisocyanate.
Yang PF, et al.
Journal of Applied Polymer Science, 123(1), 580-584 (2012)
Norman Mangner et al.
JACC. Cardiovascular imaging, 7(12), 1198-1205 (2014-10-13)
The aim of our study was to assess geometric and functional changes of the heart in obese compared with nonobese children and adolescents. Obesity in children and adolescents has increased over the past decades and is considered a strong risk

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service