Skip to Content
Merck
All Photos(1)

Key Documents

21039

Sigma-Aldrich

Calcium carbide

granulated, technical, ≥75% (gas-volumetric)

Synonym(s):

Calcium acetylide, Calcium dicarbide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaC2
CAS Number:
Molecular Weight:
64.10
Beilstein:
3909011
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

grade

technical

Quality Level

Assay

≥75% (gas-volumetric)

form

pieces

quality

granulated

particle size

0.1-1 mm

density

2.22 g/mL at 25 °C (lit.)

SMILES string

[Ca++].[C-]#[C-]

InChI

1S/C2.Ca/c1-2;/q-2;+2

InChI key

UIXRSLJINYRGFQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Calcium carbide (CaC2), also referred to as calcium acetylide, is an ionic compound. Technical grade CaC2 is a grey solid with a granular form. The material is reactive with water and forms acetylene gas and calcium hydroxide. Our technical grade material consists of ≥75% CaC2 with the remainder of the composition being calcium oxide, phosphides, sulfides, and nitrides. CaC2 is extensively utilized in the production of acetylene and calcium cyanamide. Additionally, it has been explored in producing phase change materials and as a shape-stabilized phase change material for the efficient utilization of solar energy. Furthermore, it is being investigated as an electrode material for rechargeable calcium batteries.

Application

  • NUCLEOPHILIC ADDITION OF PHOSPHINOXIDES AND ALCOHOLS TO ACETYLENE GENERATED in situ FROM CALCIUM CARBIDE: This study discusses the reactivity of acetylene generated from calcium carbide for various organic synthesis applications (Aleksandrovna, 2024).
  • Construction of 3-Methyl-2-Substituted Benzofurans and 3-Methyl-2-Substituted Benzothiophenes Using Solid Calcium Carbide as a Substitute for Gaseous …: Describes a synthetic method using solid calcium carbide in organic synthesis, highlighting its practicality and environmental benefits (Wang et al., 2024).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3 - Water-react 1

Target Organs

Respiratory system

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mirian Elizabeth Casco et al.
Nanoscale, 11(11), 4712-4718 (2019-03-07)
We report the one-pot mechanochemical synthesis of N-doped porous carbons at room temperature using a planetary ball mill. The fast reaction (5 minutes) between calcium carbide and cyanuric chloride proceeds in absence of any solvent and displays a facile bottom-up
Yingjie Li et al.
Journal of environmental sciences (China), 68, 169-176 (2018-06-18)
For the first time, a series of alkynyl carbon materials (ACMs) were prepared via the mechanochemical reaction of CaC2 with six polyhalogenated precursors, namely CCl4, C2Cl6, C2Cl4, C6Cl6, C6Br6, and C14H4Br10 (ACM-1, ACM-2, ACM-3, ACM-4, ACM-5, and ACM-6, respectively) and
H Kjuus et al.
British journal of industrial medicine, 43(4), 237-242 (1986-04-01)
The overall mortality and the incidence of cancer have been studied among male employees at a plant producing calcium carbide. The cohort was defined as all men employed at the plant for at least 18 months in the period 1953
P Boffetta et al.
Cancer causes & control : CCC, 8(3), 444-472 (1997-05-01)
Epidemiologic evidence on the relationship between polycyclic aromatic hydrocarbons (PAH) and cancer is reviewed. High occupational exposure to PAHs occurs in several industries and occupations. Covered here are aluminum production, coal gasification, coke production, iron and steel foundries, tar distillation
Xiaoguang Jiao et al.
Science in China. Series C, Life sciences, 48 Suppl 1, 26-32 (2005-08-11)
A field experiment was carried out at the Shenyang Experimental Station of Ecology (CAS) in order to study the effects of slow-release urea fertilizers high polymer-coated urea (SRU1), SRU1 mixed with dicyandiamide DCD (SRU2), and SRU1 mixed with calcium carbide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service