Skip to Content
Merck
All Photos(1)

Key Documents

202770

Sigma-Aldrich

Beryllium oxide

99.98% trace metals basis

Synonym(s):

Beryllia, Beryllium monoxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BeO
CAS Number:
Molecular Weight:
25.01
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Assay

99.98% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: beryllium

density

3.01 g/mL at 25 °C (lit.)

SMILES string

[Be]=O

InChI

1S/Be.O

InChI key

LTPBRCUWZOMYOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Beryllium oxide is a ceramic material with a unique combination of high electrical resistance and dielectric strength with high thermal conductivity. It also shows high transmission in a wide spectral range from VUV(vacuum UV) through IR. It is widely used in the field of optical devices and electronic transistors.

Application

Beryllium oxide can be used as an additive to fabricate uranium dioxide kernels(nuclear fuel) with enhanced thermal conductivity.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Carc. 1B Inhalation - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Lungs, Respiratory system

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristin J Cummings et al.
Occupational and environmental medicine, 64(2), 134-140 (2006-10-18)
A 1998 survey at a beryllium oxide ceramics manufacturing facility found that 10% of workers hired in the previous 6 years had beryllium sensitisation as determined by the beryllium lymphocyte proliferation test (BeLPT). In response, the facility implemented an enhanced
Aleksandr B Stefaniak et al.
Analytical and bioanalytical chemistry, 391(6), 2071-2077 (2008-01-29)
Owing to the absence of readily available certified particulate reference materials (RMs), most analytical methods used to determine particulate contaminant levels in workplace or other environments are validated using solution RMs, which do not assess the robustness of the digestion
Thomas J Oatts et al.
Journal of environmental monitoring : JEM, 14(2), 391-401 (2011-10-26)
Occupational sampling and analysis for multiple elements is generally approached using various approved methods from authoritative government sources such as the National Institute for Occupational Safety and Health (NIOSH), the Occupational Safety and Health Administration (OSHA) and the Environmental Protection
Caroline Muller et al.
International journal of toxicology, 30(5), 538-545 (2011-10-21)
The toxicity of 3 chemical forms of beryllium (Be) was compared in this study. A total of 160 mice equally divided into 4 groups were exposed by inhalation (nose only) for 3 consecutive weeks, 5 d/week, 6 h/d. One group
Vincent Paquette et al.
Journal of analytical toxicology, 34(9), 562-570 (2010-11-16)
Beryllium (Be) is still not well understood from a toxicological point of view, and studies that involve the determination of different Be compounds species in tissues need to be conducted. In this paper we describe the development and validation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service