Skip to Content
Merck
All Photos(1)

Key Documents

202355

Sigma-Aldrich

Poly(propylene glycol)

average Mn ~4,000

Synonym(s):

PPG, Poly(propylene oxide)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H[OCH(CH3)CH2]nOH
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
vapor pressure:
<0.01 mmHg ( 20 °C)

vapor density

>1 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

description

functionality 1.7

mol wt

average Mn ~4,000

impurities

≤0.035% Water (Karl Fischer)

viscosity

1,300 cP(25 °C)(lit.)

hydroxyl value

28 mg KOH/g

solubility

H2O: <0.01 % (w/w) at 25 °C

density

1.004 g/mL at 25 °C

SMILES string

CC(O)CO

InChI

1S/C6H14O3/c1-5(8)4-9-6(2)3-7/h5-8H,3-4H2,1-2H3

InChI key

DUFKCOQISQKSAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(propylene glycol) (PPG) is a hydrophobic polyether that is widely used in various biomedical areas such as preparation of biocompatible polyurethanes and lubricious coatings on absorbable sutures.

Application

Poly(propylene glycol) can be used:
  • As a precursor to synthesize biodegradable poly(ether-ester) networks via UV photopolymerization.
  • As a stabilizing agent in the preparation of metal nanoparticles such as gold nanoparticles.
  • As a starting material to prepare silane-terminatedpoly(urethane-urea)s (SPUR) polymers.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

445.0 °F - closed cup

Flash Point(C)

229.44 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R De Lisi et al.
Physical chemistry chemical physics : PCCP, 13(27), 12571-12577 (2011-06-15)
The study highlighted the main forces driving the formation of hydroxypropyl-cyclodextrins (HP-CDs) + poly(propylene) glycol 725 g mol(-1) inclusion complexes. The temperature parameter was chosen as the variable to modulate the hydrophobicity of the polymer, and consequently ITC experiments as
Izzuddin Zaman et al.
Nanoscale, 4(15), 4578-4586 (2012-06-19)
In spite of extensive studies conducted on carbon nanotubes and silicate layers for their polymer-based nanocomposites, the rise of graphene now provides a more promising candidate due to its exceptionally high mechanical performance and electrical and thermal conductivities. The present
F Palazzi et al.
International endodontic journal, 45(2), 129-135 (2011-09-13)
To investigate the surface tension characteristics of 5.25% sodium hypochlorite and three recently introduced sodium hypochlorite solutions, which had been modified to reduce their surface tension: Chlor-Xtra, Hypoclean A and Hypoclean B. Freshly produced MilliQ water was used as a
Mohammed Taghi Zafarani-Moattar et al.
Biotechnology progress, 28(1), 146-156 (2011-09-29)
In this work, we proposed a novel aqueous biphasic system (ABS) composed of polypropylene glycol P400 (PPG P400) and hydrophilic ionic liquids (IL), 1-alkyl-3-methylimidazolium bromide (alkyl = ethyl or butyl), forming an upper polymer-rich phase and a lower IL-rich phase
L Hong et al.
Physical review. E, Statistical, nonlinear, and soft matter physics, 83(6 Pt 1), 061508-061508 (2011-07-30)
There are various arguments and models connecting the characteristic length associated with the boson peak vibrations ξ to the length scale of dynamical heterogeneity L(het). ξ is usually defined as the ratio of the transverse sound velocity to the boson

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service