Skip to Content
Merck
All Photos(1)

Key Documents

195995

Sigma-Aldrich

2-Thiopheneacetyl chloride

98%

Synonym(s):

2-Thienylacetyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5ClOS
CAS Number:
Molecular Weight:
160.62
Beilstein:
114547
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

4 mmHg ( 83 °C)

Assay

98%

form

liquid

refractive index

n20/D 1.551 (lit.)

bp

105-106 °C/22 mmHg (lit.)
130-135 °C/90 mmHg (lit.)

density

1.303 g/mL at 25 °C (lit.)

functional group

acyl chloride

storage temp.

2-8°C

SMILES string

ClC(=O)Cc1cccs1

InChI

1S/C6H5ClOS/c7-6(8)4-5-2-1-3-9-5/h1-3H,4H2

InChI key

AJYXPNIENRLELY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Thiopheneacetyl chloride was used in the synthesis of:
  • (S)-ethyl-1-(2-thiopheneacetyl)-3-piperidinecarboxylate, nipecotate-containing immunopotentiator
  • 5-fluorouracil-cephalosporin prodrug
  • series of new N,N′-di(thiopheneacetyl)diamines derivatives
  • 6-β-(thiophen-2′-yl)acetamidomorphine

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

James M Macdougall et al.
Bioorganic & medicinal chemistry, 12(22), 5983-5990 (2004-10-23)
A series of 6-beta-arylamidomorphines was synthesized and biologically evaluated. Various aryl substituents were introduced into the arylamidomorphines to examine substituent structure-activity relationships. Competition binding assays showed that compounds 10a-h bound to the mu opioid receptor with high affinity (0.2-0.6 nM).
Development of an efficient synthesis for a nipecotate-containing immunopotentiator.
Moher ED, et al.
Organic Process Research & Development, 8(4), 593-596 (2004)
Synthesis and Biological Evaluation of N, N'-di (thiopheneacetyl) diamines Series as Antitubercular Agents.
Nora de Souza MV, et al.
Phosphorus, Sulfur, and Silicon and the Related Elements, 183(12), 2990-2997 (2008)
Ryan M Phelan et al.
Bioorganic & medicinal chemistry letters, 19(4), 1261-1263 (2009-01-27)
An efficient synthesis of a 5-fluorouracil-cephalosporin prodrug is described for use against colorectal and other cancers in antibody and gene-directed therapies. The compound shows stability in aqueous media until specifically activated by beta-lactamase (betaL). The kinetic parameters of the 5-fluorouracil-cephalosporin
Radoslaw Laufer et al.
Bioorganic & medicinal chemistry, 22(17), 4968-4997 (2014-07-22)
TTK kinase was identified by in-house siRNA screen and pursued as a tractable, novel target for cancer treatment. A screening campaign and systematic optimization, supported by computer modeling led to an indazole core with key sulfamoylphenyl and acetamido moieties at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service