Skip to Content
Merck
All Photos(1)

Key Documents

193682

Sigma-Aldrich

2-Isobutylthiazole

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11NS
CAS Number:
Molecular Weight:
141.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.495 (lit.)

bp

180 °C (lit.)

density

0.995 g/mL at 25 °C (lit.)

SMILES string

CC(C)Cc1nccs1

InChI

1S/C7H11NS/c1-6(2)5-7-8-3-4-9-7/h3-4,6H,5H2,1-2H3

InChI key

CMPVUVUNJQERIT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

DyBOBS: a dynamic biomimetic assay for odorant-binding to odor-binding protein.
Yabuki M, et al.
Chemosensory Perception, 3(2), 108-117 (2010)
B V Burger et al.
Zeitschrift fur Naturforschung. C, Journal of biosciences, 43(9-10), 731-736 (1988-09-01)
2-Isobutyl-1,3-thiazole and its 4,5-dihydro derivative were identified in the preorbital gland secretions of the grey duiker, Sylvicapra grimmia, and the red duiker, Cephalophus natalensis, but are absent from the preorbital secretion of the blue duiker, C. monticola. These two compounds
Yair Bezman et al.
Journal of agricultural and food chemistry, 51(3), 722-726 (2003-01-23)
Significant tomato matrix effects on the volatility of certain fresh tomato odorants were found. The concentrations of odorants such as (E,E)-2,4-decadienal, beta-damascenone, and beta-ionone, in crushed fresh tomato fruit obtained by solid-phase microextraction (SPME), resulting from a tomato matrix calibration
K F Christiansen et al.
Food science and technology international = Ciencia y tecnologia de los alimentos internacional, 17(2), 143-154 (2011-03-26)
A tomato flavor enhancer, 2-isobutylthiazole (IBT), was added (5 mg/kg) to dressings emulsified with either a whey protein concentrate-80 (WPC-80), a WPC-80 hydrolysate or β-lactoglobulin at high pressure (70 MPa) at either 20 or 75 °C. The short (2-4 min)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service