Skip to Content
Merck
All Photos(1)

Key Documents

192414

Sigma-Aldrich

Ethyl 2-chloropropionate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CHClCOOC2H5
CAS Number:
Molecular Weight:
136.58
Beilstein:
773906
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.417 (lit.)

bp

146-149 °C (lit.)

solubility

alcohol: miscible
diethyl ether: miscible
water: insoluble

density

1.072 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

CCOC(=O)C(C)Cl

InChI

1S/C5H9ClO2/c1-3-8-5(7)4(2)6/h4H,3H2,1-2H3

InChI key

JEAVBVKAYUCPAQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethyl 2-chloropropionate was used as initiator during the synthesis of poly(2-(diethylamino)ethyl methacrylate) and poly(N-isopropylacrylamide) via atom transfer radical polymerization. It was also used in the synthesis of end-functionalized poly(N-isopropylacrylamide) with pyrenyl group.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

100.4 °F

Flash Point(C)

38 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and thermoresponsive property of end-functionalized poly (N-isopropylacrylamide) with pyrenyl group.
Duan Q, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 44(3), 1117-1124 (2006)
Xiaoze Jiang et al.
Biomacromolecules, 8(10), 3184-3192 (2007-09-25)
A double hydrophilic ABC triblock copolymer, poly(2-(diethylamino)ethyl methacrylate)-b-poly(2-(dimethylamino)ethyl methacrylate)-b-poly(N-isopropylacrylamide) (PDEA-b-PDMA-b-PNIPAM), containing the well-known pH-responsive PDEA block and thermoresponsive PNIPAM block, was synthesized by atom transfer radical polymerization via sequential monomer addition using ethyl 2-chloropropionate as the initiator. The obtained triblock

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service