Skip to Content
Merck
All Photos(2)

Key Documents

189219

Sigma-Aldrich

Coumachlor

98%

Synonym(s):

(±)-3-(α-Acetonyl-p-chlorobenzyl)-4-hydroxycoumarin, p-Chlorowarfarin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H15ClO4
CAS Number:
Molecular Weight:
342.77
Beilstein:
6819431
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

168-170 °C (lit.)

solubility

acetone: soluble
alcohol: soluble
benzene: slightly soluble
chloroform: soluble
diethyl ether: slightly soluble
water: insoluble

functional group

chloro
ester
ketone

SMILES string

CC(=O)CC(c1ccc(Cl)cc1)C2=C(O)c3ccccc3OC2=O

InChI

1S/C19H15ClO4/c1-11(21)10-15(12-6-8-13(20)9-7-12)17-18(22)14-4-2-3-5-16(14)24-19(17)23/h2-9,15,22H,10H2,1H3

InChI key

DEKWZWCFHUABHE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Coumachlor is an anticoagulant rodenticide. It forms complex with zirconium.

Application

Coumachlor was used as internal standard for simultaneous enantioseparation of (+/-)-warfarin by chiral capillary electrochromatography with electrospray ionization mass spectrometry.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 3 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jakob Avi Shimshoni et al.
Journal of veterinary diagnostic investigation : official publication of the American Association of Veterinary Laboratory Diagnosticians, Inc, 25(6), 736-743 (2013-10-02)
Anticoagulant rodenticides are frequently a cause of poisoning of domestic animals, wildlife, and human beings. A toxicosis in 6,000 laying hens caused by the malicious addition of unknown amounts of coumatetralyl bait as well as the insecticides aldicarb, methomyl, and
Jack Zheng et al.
Analytical chemistry, 75(22), 6295-6305 (2003-11-18)
The hyphenation of chiral capillary electrochromatography (CEC) with electrospray ionization mass spectrometry (ESI-MS) is very challenging but promising due to the fact that it combines sensitivity with high specificity and selectivity. In this work, CEC capillaries packed with (3R,4S)-Whelk-O1 chiral
Jingguo Hou et al.
Journal of chromatography. A, 1159(1-2), 208-216 (2007-05-15)
Warfarin is a widely used oral anticoagulant which is mostly administrated as a racemic mixture containing equal amount of R- and S-enantiomers. The two enantiomers are shown to exhibit significant differences in pharmacokinetics and pharmacodynamics. In this study, a new
Jack Zheng et al.
Journal of chromatography. A, 1005(1-2), 177-187 (2003-08-20)
The capillary electrochromatographic separations of three acidic enantiomers (carprofen, coumachlor and warfarin) were studied on a capillary column packed with 5 microm (3R,4S)-Whelk-O 1 chiral stationary phase. The influence of several experimental parameters (mobile phase pH, type of background electrolyte
Wenjian Lao et al.
Journal of chromatography. A, 1217(42), 6545-6554 (2010-09-18)
Unusual peak profiles of warfarin were characterized on two oligoproline chiral stationary phases (CSPs). The pattern of 1st peak (S(-)) broadening and the 2nd peak (R(+)) compression was observed under mobile phase of hexane (0.1% TFA)/2-propanol (IPA) on a triproline

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service