Skip to Content
Merck
All Photos(1)

Key Documents

187844

Sigma-Aldrich

1-Bromo-3,5-dimethyladamantane

98%

Synonym(s):

1-Bromo-3,5-dimethyltricyclo[3.3.1.13,7]decane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H19Br
CAS Number:
Molecular Weight:
243.18
Beilstein:
1927514
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.52 (lit.)

bp

201 °C (lit.)

density

1.224 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

C[C@]12CC3C[C@](C)(C1)CC(Br)(C3)C2

InChI

1S/C12H19Br/c1-10-3-9-4-11(2,6-10)8-12(13,5-9)7-10/h9H,3-8H2,1-2H3/t9-,10+,11-,12-

InChI key

QUCXLVDIVQWYJR-CDECOKDKSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Bromo-3,5-dimethyladamantane was used in the one pot synthesis of 1,3-dicarbonyl adamantanes. It was also used in the synthesis of 3,5-dimethyladamantan-1-ol.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 3, 5-dimethyladamantan-1-ol by reaction of 1, 3-dimethyladamantane with bromotrichloromethane and water in the presence of manganese complexes.
Khusnutdinov RI, et al.
Russ. J. Org. Chem., 50(1), 25-28 (2014)
The first one-pot synthesis of 1, 3-dicarbonyl adamantanes from 1-bromoadamantanes.
Akhrem IS, et al.
Mendeleev Communications, 21(5), 259-261 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service