Skip to Content
Merck
All Photos(1)

Key Documents

187593

Sigma-Aldrich

Tripropylene glycol

97%

Synonym(s):

[(1-Methyl-1,2-ethanediyl)bis(oxy)]bispropanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H[OC2H3(CH3)]3OH
CAS Number:
Molecular Weight:
192.25
Beilstein:
2235421
EC Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

vapor density

6.63 (vs air)

vapor pressure

<0.01 mmHg ( 25 °C)

Assay

97%

form

liquid

refractive index

n20/D 1.444 (lit.)

bp

273 °C (lit.)

density

1.021 g/mL at 25 °C (lit.)

SMILES string

O(C(O)CC)C(COC(O)CC)C

InChI

1S/C9H20O4/c1-4-8(10)12-6-7(3)13-9(11)5-2/h7-11H,4-6H2,1-3H3

InChI key

SNAJJYBUNRAGEF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Development of New Dipropylene Glycol/Tripropylene Glycol Process: This research outlines the development of a new process for producing dipropylene glycol and tripropylene glycol, highlighting its market applications in denaturing and toughness enhancement (Yoshii et al., 2014).
  • Synthesis of silver particles on bamboo charcoal: Utilizes tripropylene glycol as a reducing agent for the synthesis of silver nanoparticles, offering potential applications in material science for creating conductive and antibacterial surfaces (Chiang & Yeh, 2014).
  • Tripropylene glycol diacrylate-based support ink: Discusses the modification of tripropylene glycol diacrylate for 3D printing applications, focusing on its use in creating support structures that can be easily removed post-manufacture (He et al., 2017).
  • Viscosity of epoxy acrylate-tripropylene glycol diacrylate mixtures: Studies the temperature dependence of viscosity in these mixtures, important for applications in ultraviolet-curable coatings, which are widely used in both material science and industrial applications (Liu et al., 2015).
  • Combustion and emissions in diesel engines: Investigates the use of tripropylene glycol as an additive in diesel fuel to improve combustion and reduce exhaust emissions, relevant for environmental chemistry and energy sector applications (Hazar, 2017).

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

293.0 °F - closed cup

Flash Point(C)

145 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chin-Ping Kung et al.
International journal of pharmaceutics, 586, 119538-119538 (2020-06-17)
Methadone appears to be a promising candidate for pain management. Previously, we conducted a comprehensive characterization study of methadone base and evaluated the dermal delivery of methadone from various neat solvents. Four solvents, namely d-limonene (LIM), ethyl oleate (EO), Transcutol®

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service