Skip to Content
Merck
All Photos(1)

Key Documents

186384

Sigma-Aldrich

N,N-Dimethyl-p-phenylenediamine sulfate salt

98%

Synonym(s):

4-(Dimethylamino)aniline sulfate salt, 4-Amino-N,N-dimethylaniline sulfate salt, DMPPDA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NC6H4NH2·H2SO4
CAS Number:
Molecular Weight:
234.27
Beilstein:
4612278
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

495 °C (lit.)

mp

200-205 °C (dec.) (lit.)

solubility

water: soluble 50 mg/mL, clear to hazy, colorless to faintly yellow or pink

functional group

amine

SMILES string

OS(O)(=O)=O.CN(C)c1ccc(N)cc1

InChI

1S/C8H12N2.H2O4S/c1-10(2)8-5-3-7(9)4-6-8;1-5(2,3)4/h3-6H,9H2,1-2H3;(H2,1,2,3,4)

InChI key

GLUKPDKNLKRLHX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N-Dimethyl-p-phenylenediamine sulfate salt was used:
  • to investigate the role of H2S in cardioprotection induced by ischemic preconditioning in rat heart
  • in spectrophotometric determination of gold(III) in gold-pharmaceuticals such as sodium aurothimaleate and auranofin
  • to investigate colonic tissue H2S production using the acetate trapping assay system

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Kyle L Flannigan et al.
American journal of physiology. Gastrointestinal and liver physiology, 301(1), G188-G193 (2011-04-09)
Hydrogen sulfide (H(2)S) is an important modulator of many aspects of digestive function, both in health and disease. Colonic tissue H(2)S synthesis increases markedly during injury and inflammation and appears to contribute to resolution. Some of the bacteria residing in
Jin-Song Bian et al.
The Journal of pharmacology and experimental therapeutics, 316(2), 670-678 (2005-10-06)
Endogenous H(2)S is synthesized mainly by cystathionine gamma-lyase in the heart. The present study investigated the role of H(2)S in cardioprotection induced by ischemic preconditioning. We have examined the effect of endogenous H(2)S and exogenous application of NaHS (H(2)S donor)
New and sensitive spectrophotometric determination of gold (III) with N, N-dimethyl-p-phenylenediamine and potassium persulfate.
Mori I, et al.
Analytical Letters, 30(5), 953-961 (1997)
Claudineia R Silva et al.
Talanta, 85(3), 1703-1705 (2011-08-03)
A spectrophotometric flow injection procedure involving N,N-dimethyl-p-phenylenediamine (DMPD) is applied to the sulfide monitoring of a sugar fermentation by Saccharomyces cerevisiae under laboratory conditions. The gaseous chemical species evolving from the fermentative process, mainly CO(2), are trapped allowing a cleaned
V Leskovac et al.
The Italian journal of biochemistry, 45(1), 9-18 (1996-03-01)
The steady-state kinetics, product identification, stoichiometries, and solvent isotope effects of yeast alcohol dehydrogenase catalyzed reduction of p-nitroso-N,N-dimethylaniline (NDMA) by NADH, are reported. NDMA is enzymatically reduced to p-hydroxylamine-N,N-dimethylaniline, which is further enzymatically dehydrated to corresponding quinonediimine cation (QDI+). QDI+

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service