Skip to Content
Merck
All Photos(1)

Key Documents

185760

Sigma-Aldrich

1-Chloro-2-nitrobenzene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4NO2
CAS Number:
Molecular Weight:
157.55
Beilstein:
509273
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

5.4 (vs air)

Quality Level

vapor pressure

0.04 mmHg ( 25 °C)

Assay

99%

form

solid

autoignition temp.

500 °F

expl. lim.

8.7 %

bp

246 °C (lit.)

mp

31-33 °C (lit.)

solubility

alcohol: soluble
benzene: soluble
diethyl ether: soluble
water: insoluble

density

1.348 g/mL at 25 °C (lit.)

functional group

chloro
nitro

SMILES string

[O-][N+](=O)c1ccccc1Cl

InChI

1S/C6H4ClNO2/c7-5-3-1-2-4-6(5)8(9)10/h1-4H

InChI key

BFCFYVKQTRLZHA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Chloro-2-nitrobenzene was reduced to o-chloroaniline by using 5% platinum on carbon with ammonium formale or forinic acid as hydrogen donor. Reaction of 1-chloro-2-nitrobenzene with dinitrogen pentoxide in dichloromethane is strongly catalysed by H-Faujasite-720(zeolite).

Application

1-Chloro-2-nitrobenzene was used in the synthesis of 1-hydroxybenzotriazole derivatives.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

258.8 °F - closed cup

Flash Point(C)

126 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Catalytic transfer hydrogenation of aromatic nitro compounds by employing ammonium formate and 5% platinum on carbon.
Gowda DC and Mahesh B.
Synthetic Communications, 30(20), 3639-3644 (2000)
Zeolite catalysis of aromatic nitrations with dinitrogen pentoxide.
Claridge RP, et al.
J. Chem. Soc. Perkin Trans. II, 9, 1815-1818 (1999)
Spectrophotometric Determination of pKa's of 1-Hydroxybenzotriazole and Oxime Derivatives in 95% Acetonitrile-Water.
Fathalla MF and Khattab SN.
J. Chem. Soc. Pak., 33(3), 324-324 (2011)
Haizhen Wu et al.
Journal of environmental sciences (China), 21(1), 89-95 (2009-05-01)
A bacterial strain that utilized o-chloronitrobenzene (o-CNB) as the sole carbon, nitrogen and energy sources was isolated from an activated sludge collected from an industrial waste treatment plant. It was identified as Pseudomonas putida based on its morphology, physiological, and
Hong Liu et al.
Applied and environmental microbiology, 77(13), 4547-4552 (2011-05-24)
Pseudomonas stutzeri ZWLR2-1 utilizes 2-chloronitrobenzene (2CNB) as a sole source of carbon, nitrogen, and energy. To identify genes involved in this pathway, a 16.2-kb DNA fragment containing putative 2CNB dioxygenase genes was cloned and sequenced. Of the products from the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service