185442
Hexachloroethane
99%
Synonym(s):
Perchloroethane
About This Item
Recommended Products
vapor density
8.16 (vs air)
vapor pressure
0.4 mmHg ( 20 °C)
Assay
99%
form
solid
mp
183-185 °C (dec.) (lit.)
solubility
alcohol: soluble
benzene: soluble
chloroform: soluble
diethyl ether: soluble
oil: soluble
water: insoluble
density
2.091 g/mL at 25 °C (lit.)
functional group
chloro
SMILES string
ClC(Cl)(Cl)C(Cl)(Cl)Cl
InChI
1S/C2Cl6/c3-1(4,5)2(6,7)8
InChI key
VHHHONWQHHHLTI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service