Skip to Content
Merck
All Photos(2)

Key Documents

182427

Sigma-Aldrich

Polystyrene

average Mw ~280,000 by GPC

Synonym(s):

PS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(C6H5)]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

beads or pellets

Quality Level

mol wt

average Mw ~280,000 by GPC

refractive index

n20/D 1.5916

transition temp

Tg 100 °C

density

1.047 g/mL at 25 °C

SMILES string

c1(ccccc1)C=C

InChI

1S/C8H8/c1-2-8-6-4-3-5-7-8/h2-7H,1H2

InChI key

PPBRXRYQALVLMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Polystyrene is a high molecular weight aromatic synthetic polymer madefrom the polymerization of styrene. It is a hydrophobic durable thermoplasticwidely used in food containers, packaging foam, and construction materials dueto its good mechanical properties and low cost.

Application

Polystyrene can be added to nanocatalyst suspension to prepare a polystyrene-photocatalyst thick film. This film can be used as a photoanode in an airlift photocatalytic fuel cell (APFC).

Polystyrene can be used as a precursor to prepare graphene film and 3D graphene sponges by pyrolysis under argon atmosphere. The filmogenic property of polystyrene can be exploited to prepare 3D graphene sponges with uniform pore sizes.

It can also be used to prepare dyed polystyrene (PS) film for routine dosimetry in radiation processing. Dyed PS film has high post-irradiation stability.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Alejandra Rendón-Patiño et al.
Nanomaterials (Basel, Switzerland), 9(1) (2019-01-19)
Polystyrene as a thin film on arbitrary substrates or pellets form defective graphene/graphitic films or powders that can be dispersed in water and organic solvents. The materials were characterized by visible absorption, Raman and X-ray photoelectron spectroscopy, electron and atomic
Ba Thanh Ho et al.
Critical reviews in biotechnology, 38(2), 308-320 (2017-08-03)
Polystyrene is a widely used plastic in many aspects of human life and in industries due to its useful characteristics of low cost, light weight, ease of manufacture, versatility, thermal efficiency, durability, and moisture resistance. However, polystyrene is very stable
J K R Weber et al.
The Review of scientific instruments, 80(8), 083904-083904 (2009-09-04)
A single-axis acoustic levitator was constructed and used to levitate liquid and solid drops of 1-3 mm in diameter at temperatures in the range -40 to +40 degrees C. The levitator comprised (i) two acoustic transducers mounted on a rigid
Kevin Kendall et al.
Nanotechnology, 20(27), 275701-275701 (2009-06-18)
The weak molecular attractions of nanoparticles are important because they drive self-assembly mechanisms, allow processing in dispersions e.g. of pigments, catalysts or device structures, influence disease through the attraction of viruses to cells and also cause potential toxic effects through
Di Wei et al.
ACS nano, 2(12), 2526-2530 (2009-02-12)
We have fabricated single-walled carbon nanotube (SWNT) field effect transistors (FETs) with molybdenum source and drain contacts. Normally, these devices operate only as p-channel transistors, however, after polystyrene latex nanospheres were attached to the nanotubes close to the contacts, they

Questions

1–4 of 4 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. What is the tacticity of this PS? Is it atactic?

    1 answer
    1. This product is atactic.

      Helpful?

  4. Hello, I'd like to know how these polystyrene samples are typically synthesized? (Free radical polymerization? Anionic living polymerization?) Thanks, Stanley

    1 answer
    1. Unfortunately, the method of polymerization is considered proprietary.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service