181501
Poly(1,4-butylene adipate)
average Mw ~12,000 by GPC
Synonym(s):
Adipic acid-1,4-butanediol copolymer, PBAG, Poly(butylene adipate), Poly(tetramethylene adipate)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
[O(CH2)4O2C(CH2)4CO]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
form
chips
mol wt
average Mw ~12,000 by GPC
transition temp
Tg −68 °C
Tm 54 °C
density
1.019 g/mL at 25 °C (lit.)
SMILES string
OCCCCO.OC(=O)CCCCC(O)=O
InChI
1S/C6H10O4.C4H10O2/c7-5(8)3-1-2-4-6(9)10;5-3-1-2-4-6/h1-4H2,(H,7,8)(H,9,10);5-6H,1-4H2
InChI key
RNSLCHIAOHUARI-UHFFFAOYSA-N
Application
Injection molding for automotive, mechanical and electronic industries.
Features and Benefits
Excellent heat resistance, low creep and high stiffness at elevated temperatures.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service