Skip to Content
Merck
All Photos(1)

Key Documents

178314

Sigma-Aldrich

N-Aminophthalimide

technical grade, 90%

Synonym(s):

N,N-Phthaloylhydrazine, unsym.

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6N2O2
CAS Number:
Molecular Weight:
162.15
Beilstein:
383756
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

form

powder

mp

200-202 °C (lit.)

functional group

imide

SMILES string

NN1C(=O)c2ccccc2C1=O

InChI

1S/C8H6N2O2/c9-10-7(11)5-3-1-2-4-6(5)8(10)12/h1-4H,9H2

InChI key

KSILMCDYDAKOJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-Aminophthalimide was employed in the aziridination of chiral N-enoyl sultams. It was also used in the synthesis of n-phthalimidoaziridines.

Other Notes

Remainder phthalhydrazide

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the Chemical Society. Chemical Communications, 1074-1074 (1993)
Kung-Shou Yang et al.
Organic letters, 4(7), 1107-1109 (2002-04-02)
[reaction: see text] Reaction of various N-enoyl oxazolidinones 5a-f with N-aminophthalimide and lead tetraacetate in the presence of camphor-derived chiral ligands provides the desired N-phthalimidoaziridines 6a-f in good to high enantiomeric excess (67-95% ee) at 0 degrees C within 15

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service