177296
4,4′-Dipyridyl hydrate
98%
Synonym(s):
4,4′-Bipyridine
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C10H8N2 · xH2O
CAS Number:
Molecular Weight:
156.18 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
solid
bp
304.8 °C (lit.)
mp
70-74 °C (lit.)
SMILES string
[H]O[H].c1cc(ccn1)-c2ccncc2
InChI
1S/C10H8N2.H2O/c1-5-11-6-2-9(1)10-3-7-12-8-4-10;/h1-8H;1H2
InChI key
JJRKHJXMBCYHSH-UHFFFAOYSA-N
Application
4,4′-Dipyridyl hydrate was used in the preparation of hydrogen-bonded coordination polymers.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Novel hydrogen-bonded two-and three-dimensional networks generated from the reaction of metal nitrate hydrates (M= Cd, Co) with the bidentate linear ligand 4, 4′-bipyridine.
J. Chem. Soc., Dalton Trans., 5, 775-780 (2000)
Bing Liu et al.
Molecules (Basel, Switzerland), 20(7), 12341-12363 (2015-07-23)
The synthesis and structural characterization of five transition metal complexes with different dimensionality and incorporating residues of 3-amino-1H-1,2,4-triazole-5-carboxylic acid (H2atrc) is reported: [Zn(Hatrc)2(H2O)] (1), [Mn(Hatrc)2(H2O)2]·2H2O (2), [Fe2(Hatrc)4(OH)2]·6H2O (3), [Cd(Hatrc)2(H2O)]n (4), and [Mn(atrc)(H2O)]n·nH2O (5). These materials could be prepared from solution
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service