Skip to Content
Merck
All Photos(1)

Key Documents

176877

Sigma-Aldrich

Phthalan

97%

Synonym(s):

o-Xylylene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8O
CAS Number:
Molecular Weight:
120.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.546 (lit.)

bp

192 °C (lit.)

density

1.098 g/mL at 25 °C (lit.)

functional group

ether

SMILES string

C1OCc2ccccc12

InChI

1S/C8H8O/c1-2-4-8-6-9-5-7(8)3-1/h1-4H,5-6H2

InChI key

SFLGSKRGOWRGBR-UHFFFAOYSA-N

Application

Phthalan was used in the synthesis of functionalised benzylic alcohols.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

1, 2-Di (lithiomethyl) benzene from phthalan: sequential introduction of two different electrophiles.
Almena J, et al.
Tetrahedron, 51(11), 3351-3364 (1995)
Matheus Henrique Reis da Silva et al.
Archives of microbiology, 202(10), 2779-2789 (2020-08-04)
Endophytes are considered one of the most important microbial resources for obtaining biomolecules of therapeutic use. Passiflora incarnata, widely employed by the pharmaceutical industry, shows therapeutic effects on anxiety, nervousness, constipation, dyspepsia and insomnia based on their antioxidant compounds. In
Jingyong Liu et al.
Bioresource technology, 277, 104-116 (2019-01-20)
(Co-)pyrolysis behaviors were quantified using TG and Py-GC/MS analyses as a function of the two fuels of sewage sludge (SS) and water hyacinth (WH), five atmospheres, six blend ratios, and three heating rates. Co-pyrolysis performance, gaseous characterizations and optimization analyses
Giuliana Donadio et al.
PloS one, 10(4), e0124427-e0124427 (2015-04-29)
Monocyclic phenols and catechols are important antioxidant compounds for the food and pharmaceutic industries; their production through biotransformation of low-added value starting compounds is of major biotechnological interest. The toluene o-xylene monooxygenase (ToMO) from Pseudomonas sp. OX1 is a bacterial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service