Skip to Content
Merck
All Photos(1)

Key Documents

176230

Sigma-Aldrich

Triethyloxonium tetrafluoroborate solution

1.0 M in methylene chloride

Synonym(s):

Et3OBF4, Meerwein′s reagent, Triethyloxonium fluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3O(BF4)
CAS Number:
Molecular Weight:
189.99
Beilstein:
3598090
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

concentration

1.0 M in methylene chloride

density

1.328 g/mL at 25 °C

functional group

ether

storage temp.

2-8°C

SMILES string

F[B-](F)(F)F.CC[O+](CC)CC

InChI

1S/C6H15O.BF4/c1-4-7(5-2)6-3;2-1(3,4)5/h4-6H2,1-3H3;/q+1;-1

InChI key

IYDQMLLDOVRSJJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in the preparation of ω-aminoesters from lactams.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthetic Communications, 18, 1625-1625 (1988)
J Aguado et al.
Biotechnology and applied biochemistry, 17 ( Pt 1), 49-55 (1993-02-01)
beta-Glucosidase from Penicillium funiculosum was immobilized on nylon powder previously activated with triethyloxonium tetrafluoroborate, 1,2-diaminoethane and glutaraldehyde. The activation of the nylon powder and the immobilization processes were studied and optimized for the enzyme and the matrix. A high activity
Some detoxification methods for N-nitrosamine-contaminated wastes.
M Castegnaro et al.
IARC scientific publications, (41)(41), 151-157 (1982-01-01)
R Ben Avraham et al.
FEBS letters, 180(2), 239-242 (1985-01-28)
Treatment of trypsin with triethyloxonium tetrafluoroborate at pH 8, 25 degrees C, results in abolition of binding to the enzyme of specific cationic substrates and inhibitors. The binding constant of soybean trypsin inhibitor to ethylated trypsin is 10000-fold smaller than
Enea Pagliano et al.
Analytical chemistry, 84(5), 2592-2596 (2012-02-11)
The alkylation of nitrite and nitrate by triethyloxonium tetrafluoroborate allows determination of their ethyl esters by headspace gas chromatography/mass spectrometry (GC/MS). In the present study, significant improvement in analytical performance is achieved using negative chemical ionization providing detection limits of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service