175102
Fluoroantimonic acid
purified by triple-distillation
Synonym(s):
Fluoroantimonic acid, Hexafluoroantimonic acid, Hexafluorostibonic acid, Monohydrogen hexafluoroantimonate
About This Item
Recommended Products
vapor pressure
14 mmHg ( 18 °C)
Quality Level
form
liquid
purified by
triple-distillation
density
2.885 g/mL at 25 °C (lit.)
SMILES string
F.F[Sb](F)(F)(F)F
InChI
1S/6FH.Sb/h6*1H;/q;;;;;;+5/p-5
InChI key
MBAKFIZHTUAVJN-UHFFFAOYSA-I
Related Categories
General description
Application
Packaging
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1A
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service