Skip to Content
Merck
All Photos(1)

Key Documents

167045

Sigma-Aldrich

Methyl 3-hydroxy-2-naphthoate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC10H6CO2CH3
CAS Number:
Molecular Weight:
202.21
Beilstein:
1074526
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

bp

205-207 °C (lit.)

mp

73-75 °C (lit.)

functional group

ester

SMILES string

COC(=O)c1cc2ccccc2cc1O

InChI

1S/C12H10O3/c1-15-12(14)10-6-8-4-2-3-5-9(8)7-11(10)13/h2-7,13H,1H3

InChI key

YVVBECLPRBAATK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl 3-hydroxy-2-naphthoate undergoes asymmetric oxidative coupling using mono-N-alkylated octahydrobinaphthyl-2,2′-diamine chiral ligands.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective oxidative coupling of methyl 3-hydroxy-2-naphthoate using mono-< i> N</i>-alkylated octahydrobinaphthyl-2, 2'-diamine ligand.
Kim KH, et al.
Tetrahedron, 60(41), 9037-9042 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service