Skip to Content
Merck
All Photos(1)

Key Documents

163155

Sigma-Aldrich

Ethyl trichloroacetate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3CCO2CH2CH3
CAS Number:
Molecular Weight:
191.44
Beilstein:
1761567
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.453 (lit.)

bp

168 °C (lit.)

solubility

alcohol: miscible(lit.)
diethyl ether: miscible(lit.)
water: insoluble(lit.)

density

1.378 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C(Cl)(Cl)Cl

InChI

1S/C4H5Cl3O2/c1-2-9-3(8)4(5,6)7/h2H2,1H3

InChI key

SJMLNDPIJZBEKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl trichloroacetate undergoes atom-transfer radical addition reaction with styrene catalyzed by the Ru-pentamethylcyclopentadienyl complexes.

Application

Ethyl trichloroacetate was used in the synthesis of dichlorocarbon precursor, phenyl(trichloromethyl)methyl.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An Improved Synthesis of Phenyl (trichloro-methyl) mercury from Sodium Methoxide and Ethyl Trichloroacetate.
Schweizer EE and O'Neill GJ.
The Journal of Organic Chemistry, 28(3), 851-852 (1963)
Mariano A Fernández-Zúmel et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 15(43), 11601-11607 (2009-09-15)
Kinetic and spectroscopic analyses were performed to gain information about the mechanism of atom-transfer radical reactions catalyzed by the complexes [RuCl2Cp*(PPh3)] and [RuClCp*(PPh3)2] (Cp*=pentamethylcyclopentadienyl), in the presence and in the absence of the reducing agent magnesium. The reactions of styrene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service