Skip to Content
Merck
All Photos(3)

Key Documents

155225

Sigma-Aldrich

2-Piperidinemethanol

97%

Synonym(s):

2-(Hydroxymethyl)piperidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO
CAS Number:
Molecular Weight:
115.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

68-70 °C (lit.)

functional group

hydroxyl

SMILES string

OCC1CCCCN1

InChI

1S/C6H13NO/c8-5-6-3-1-2-4-7-6/h6-8H,1-5H2

InChI key

PRAYXGYYVXRDDW-UHFFFAOYSA-N

Application

2-Piperidinemethanol was used in the synthesis of 3-phenyloctahydropyrido[2,1-c][1,4]oxazine hydrochloride and the 10R and 10S diastereomers.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

T N Riley et al.
Journal of medicinal chemistry, 19(2), 334-336 (1976-02-01)
3-Phenyloctahydropyrido[2,1-c][1,4]oxazine hydrochloride and the 10R and 10S diastereomers have been synthesized from (+/-)-, (+)-, and (-)-2-piperidinemethanol. Treatment of 2-piperidinemethanol with alpha-bromoacetophenone gave 3-hydroxy-3-phenyloctahydropyrido[2,1-c][1,4]oxazine which was readily converted to the 3-phenyl derivative by catalytic hydrogenolysis. These compounds were shown to possess
Mostafa Hadei et al.
Journal of occupational medicine and toxicology (London, England), 13, 30-30 (2018-10-03)
The use of cosmetic products in beauty salons emits numerous kinds of toxic air pollutants. The objectives of this study were to measure the concentrations of benzene, toluene, ethylbenzene, xylene, formaldehyde, and acetaldehyde in 20 large beauty salons in Tehran
Daniel C O'Brien et al.
Otolaryngology--head and neck surgery : official journal of American Academy of Otolaryngology-Head and Neck Surgery, 163(3), 508-516 (2020-05-27)
To assess the exposure of surgical personnel to known carcinogens during pediatric tonsillectomy and adenoidectomy (T&A) and compare the efficacy of surgical smoke evacuation systems during T&A. Prospective, case series. Tertiary children's hospital. The present study assessed operating room workers'

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service