Skip to Content
Merck
All Photos(2)

Key Documents

154288

Sigma-Aldrich

4-Methoxy-6-methyl-2H-pyran-2-one

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8O3
CAS Number:
Molecular Weight:
140.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

form

solid

mp

83-86 °C (lit.)

functional group

ester
ether

SMILES string

COC1=CC(=O)OC(C)=C1

InChI

1S/C7H8O3/c1-5-3-6(9-2)4-7(8)10-5/h3-4H,1-2H3

InChI key

MTZAUZNQAMNFME-UHFFFAOYSA-N

Application

4-Methoxy-6-methyl-2H-pyran-2-one was used as a model compound to study the assignment of ring proton resonances in 4-methoxy-6-nonyl-2H-pyran-2-one[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pyrones. VI. The total synthesis of phacidin.
Canadian Journal of Chemistry, 58(20), 2158-2160 (1980)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service