Skip to Content
Merck
All Photos(1)

Key Documents

15402

Sigma-Aldrich

Boc-Dap-OH

≥98.0% (TLC)

Synonym(s):

(S)-2-[(tert-Butoxycarbonyl)amino]-3-aminopropionic acid, (S)-3-Amino-2-(tert-butoxycarbonyl)aminopropionic acid, (S)-3-Amino-2-(tert-butoxycarbonylamino)propanoic acid, 3-Amino-(tert-butoxycarbonyl)-L-alanine, Nα-BOC-(S)-β-aminoalanine, Nα-Boc-L-β-aminoalanine, N2-(tert-Butoxycarbonyl)-(S)-2,3-diaminopropionic acid, N2-tert-Butoxycarbonyl-L-2,3-diaminopropionic acid, Nα-Boc-L-2,3-diaminopropionic acid, Boc-Dpr-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16N2O4
CAS Number:
Molecular Weight:
204.22
Beilstein:
4182136
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (TLC)

form

solid

optical activity

[α]20/D +5.5±1°, c = 1% in methanol: water (1:1)

reaction suitability

reaction type: Boc solid-phase peptide synthesis

impurities

~3% water

mp

210 °C (dec.)

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N[C@@H](CN)C(O)=O

InChI

1S/C8H16N2O4/c1-8(2,3)14-7(13)10-5(4-9)6(11)12/h5H,4,9H2,1-3H3,(H,10,13)(H,11,12)/t5-/m0/s1

InChI key

KRJLRVZLNABMAT-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Protein assembly directed by synthetic molecular recognition motifs
  • Solid phase synthesis of gramicidin S cyclic analogs with antibiotic and hemolytic activities
  • Synthesis of HCV protease inhibitor modified analogs
  • Solid phase synthesis of peptidic V1a receptor agonists
  • Directed peptide assembly at lipid-water interface

Other Notes

Monoprotected derivative of DAP; used e.g. in the synthesis of glucosamine synthase inhibitors, a myosin kinase inhibitor. Preparation of peptides with metal complexing groups.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Andruszkiewicz et al.
Journal of medicinal chemistry, 33(10), 2755-2759 (1990-10-01)
Six peptide conjugates consisting of either norvaline, methionine, or lysine and N3-(iodoacetyl)-L-2,3-diaminopropanoic acid--a strong, irreversible inactivator of bacterial and fungal glucosamine-6-phosphate synthase--were synthesized and their antibacterial and antifungal activities were evaluated. Antimicrobial potencies of these peptides were correlated with their
F. Ruan et al.
The Journal of Organic Chemistry, 56, 4347-4347 (1991)
N Kucharczyk et al.
Biochemistry, 29(15), 3668-3676 (1990-04-17)
A mechanistic investigation of the inactivation of Escherichia coli glucosamine-6-phosphate synthase by N3-(4-methoxyfumaroyl)-L-2,3-diaminopropionate (FMDP) was undertaken. On the basis of the known participation of the N-terminal cysteine residue in this process [Chmara et al. (1986) Biochim. Biophys. Acta 870, 357;
J T Hunt et al.
The Biochemical journal, 257(1), 73-78 (1989-01-01)
Although the amino acid residues that are important for peptide substrates of myosin light-chain kinase have been reported, those that are important for peptide inhibitors of this enzyme have not previously been investigated. Synthetic peptides based on the sequence Lys11-Lys12-Arg13-Ala-Ala-Arg16-Ala-Thr-Ser19

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service