Skip to Content
Merck
All Photos(1)

Key Documents

153664

Sigma-Aldrich

2,4,6-Triphenoxy-1,3,5-triazine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H15N3O3
CAS Number:
Molecular Weight:
357.36
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

232-235 °C (lit.)

functional group

phenoxy

SMILES string

O(c1ccccc1)c2nc(Oc3ccccc3)nc(Oc4ccccc4)n2

InChI

1S/C21H15N3O3/c1-4-10-16(11-5-1)25-19-22-20(26-17-12-6-2-7-13-17)24-21(23-19)27-18-14-8-3-9-15-18/h1-15H

InChI key

IYDYVVVAQKFGBS-UHFFFAOYSA-N

Application

2,4,6-Triphenoxy-1,3,5-triazine was used to study the steric structure effects in the triazine system for formation of polymeric structures.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carbon and nitrogen coordinated silver (I) polymers of 2, 4, 6-triphenoxy-1, 3, 5-triazine.
Munakata M, et al.
Polyhedron, 20(18), 2321-2327 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service