15302
4-(Boc-amino)-1-butanol
≥98.0% (GC)
Synonym(s):
tert-Butyl N-(4-hydroxybutyl)carbamate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
Assay
≥98.0% (GC)
reaction suitability
reagent type: cross-linking reagent
density
1.02 g/mL at 20 °C (lit.)
functional group
Boc
amine
hydroxyl
storage temp.
2-8°C
SMILES string
OCCCCNC(OC(C)(C)C)=O
InChI
1S/C9H19NO3/c1-9(2,3)13-8(12)10-6-4-5-7-11/h11H,4-7H2,1-3H3,(H,10,12)
InChI key
LIYMTLVBAVHPBU-UHFFFAOYSA-N
Application
Building block for spermidine analogues
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
J. Chem. Res. Synop., 366-366 (1996)
International journal of pharmaceutics, 561, 43-46 (2019-02-18)
Oral Thin Film (OTF) is a newly emerging drug delivery system which has many benefits for patients. Although there has been some formulation of OTF products, these have mainly been as confectionary or dental health products. The most significant benefit
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service