Skip to Content
Merck
All Photos(2)

Key Documents

15264

Sigma-Aldrich

Bis(trimethylsilylmethyl) sulfide

≥98.0%

Synonym(s):

Thiobis(methylene)bis(trimethylsilane)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H22SSi2
CAS Number:
Molecular Weight:
206.50
Beilstein:
1740046
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0%

refractive index

n20/D 1.455

bp

88 °C/21 mmHg (lit.)

density

0.844 g/mL at 20 °C (lit.)

functional group

thioether

SMILES string

C[Si](C)(C)CSC[Si](C)(C)C

InChI

1S/C8H22SSi2/c1-10(2,3)7-9-8-11(4,5)6/h7-8H2,1-6H3

InChI key

GISUCMFTNKRCPF-UHFFFAOYSA-N

Related Categories

Other Notes

Synthetic equivalent of (methylthio)methyl anion in F--promoted reactions

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A. Hosomi et al.
Synlett, 557-557 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service