Skip to Content
Merck
All Photos(1)

Key Documents

152129

Sigma-Aldrich

4-Methoxy-3-methylbenzaldehyde

99%

Synonym(s):

3-Methyl-p-anisaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(OCH3)CHO
CAS Number:
Molecular Weight:
150.17
Beilstein:
2084045
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.569 (lit.)

bp

80-85 °C/1 mmHg (lit.)

density

1.025 g/mL at 25 °C (lit.)

functional group

aldehyde

SMILES string

[H]C(=O)c1ccc(OC)c(C)c1

InChI

1S/C9H10O2/c1-7-5-8(6-10)3-4-9(7)11-2/h3-6H,1-2H3

InChI key

MYLBIQHZWFWSMH-UHFFFAOYSA-N

Application

4-Methoxy-3-methylbenzaldehyde was used in the preparation of (4-methoxy-3-methylphenyl)-2-nitroalkene.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tamer Awad et al.
Journal of chromatographic science, 45(8), 477-485 (2007-11-21)
The five side-chain regioisomers of 4-methoxy-3-methylphenethylamine constitute a unique set of compounds having an isobaric relationship with the controlled drug substance 3,4-methylenedioxymethamphetamine (3,4-MDMA or Ecstasy). These isomeric forms of the 4-methoxy-3-methylphenethylamines have mass spectra essentially equivalent to 3,4-MDMA, and all

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service