Skip to Content
Merck
All Photos(2)

Key Documents

150045

Sigma-Aldrich

2,4-Dimethyl-3-pentanol

99%

Synonym(s):

Diisopropylcarbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH(OH)CH(CH3)2
CAS Number:
Molecular Weight:
116.20
Beilstein:
1731593
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

refractive index

n20/D 1.425 (lit.)

bp

139-140 °C (lit.)

density

0.829 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CC(C)C(O)C(C)C

InChI

1S/C7H16O/c1-5(2)7(8)6(3)4/h5-8H,1-4H3

InChI key

BAYAKMPRFGNNFW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,4-Dimethyl-3-pentanol was used as a proton source in the diastereoselective coupling with 2-substituted acrylate derivatives. It was used in the polymerization of 1,1′-(1,3-phenylene) diethanol (1,3-diol) and diisopropyl adipate.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

98.6 °F - closed cup

Flash Point(C)

37 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Bart A C van As et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 13(29), 8325-8332 (2007-07-31)
The well-known dynamic kinetic resolution of secondary alcohols and esters was extended to secondary diols and diesters to afford chiral polyesters. This process is an example of iterative tandem catalysis (ITC), a polymerization method where the concurrent action of two
Hegui Gong et al.
Organic letters, 11(4), 879-882 (2009-01-28)
A mild, stereoselective method for the Ni-catalyzed synthesis of alpha-C-alkylglycosides is reported. This approach entails the reductive coupling of glycosyl bromides with activated alkenes at room temperature, with low alkene loading as an important feature. Diastereoselective coupling with 2-substituted acrylate
Liping Wang et al.
Pesticide biochemistry and physiology, 139, 46-52 (2017-06-10)
Camptothecin (CPT), a quinolone alkaloid extracted from Camptotheca acuminata Decne, exhibits potential insecticidal activities against various insect species. Our previous studies have showed that CPT induced apoptosis in Spodoptera exigua Hübner cell line and inhibited the relaxation activity of topoisomerase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service