Skip to Content
Merck
All Photos(2)

Key Documents

149799

Sigma-Aldrich

1,2,3,4-Tetraphenylnaphthalene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H4(C6H5)4
CAS Number:
Molecular Weight:
432.55
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

199-201 °C (lit.)

functional group

phenyl

SMILES string

c1ccc(cc1)-c2c(-c3ccccc3)c(-c4ccccc4)c5ccccc5c2-c6ccccc6

InChI

1S/C34H24/c1-5-15-25(16-6-1)31-29-23-13-14-24-30(29)32(26-17-7-2-8-18-26)34(28-21-11-4-12-22-28)33(31)27-19-9-3-10-20-27/h1-24H

InChI key

UCTTYTFENYGAPP-UHFFFAOYSA-N

Application

The triplet excited-state of 1,2,3,4-tetraphenylnaphthalene is a primary intermediate of the photochemical transformation of 7,7′-dimethylgerma-1,4,5,6-tetraphenyl-2,3-benzo-norbornadiene (GNB) in hexane solution.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Margarita V Kaletina et al.
The journal of physical chemistry. A, 110(50), 13341-13348 (2006-12-15)
Laser pulse photolysis experiments have shown that the triplet excited-state of 1,2,3,4-tetraphenylnaphthalene (TPN) is one of the primary intermediates of the photochemical transformation of 7,7'-dimethylgerma-1,4,5,6-tetraphenyl-2,3-benzo-norbornadiene (GNB) in hexane solution. The molar absorption of T-T absorption and the quantum yield of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service