Skip to Content
Merck
All Photos(2)

Key Documents

147494

Sigma-Aldrich

2-Ethoxybenzoic acid

98%

Synonym(s):

O-Ethylsalicylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OC6H4CO2H
CAS Number:
Molecular Weight:
166.17
Beilstein:
2691039
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

refractive index

n20/D 1.54 (lit.)

bp

174-176 °C/15 mmHg (lit.)

mp

19.3-19.5 °C (lit.)

solubility

95% ethanol: soluble 5%, clear, colorless to yellow

density

1.105 g/mL at 25 °C (lit.)

functional group

carboxylic acid

SMILES string

CCOc1ccccc1C(O)=O

InChI

1S/C9H10O3/c1-2-12-8-6-4-3-5-7(8)9(10)11/h3-6H,2H2,1H3,(H,10,11)

InChI key

XDZMPRGFOOFSBL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Ethoxybenzoic acid is dental cement. It is root-end filling material and prevents the entry of root-canal pathogens into periapical regions.

Application

2-Ethoxybenzoic acid was used in thermally induced solvent-free reactions of Ph3Bi with a series of thiols and carboxylic acids.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

209.5 °F - closed cup

Flash Point(C)

98.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

S Fujisawa et al.
Journal of dental research, 60(4), 860-864 (1981-04-01)
The binding constants (log K) of eugenol, o-ethoxy-benzoic acid, and their analogues, plus other organic compounds widely used in dentistry with bovine serum albumin, were examined with respect to the partition coefficients of these compounds in octanol-water (log P). Linear
Grazyna Bieniek et al.
Journal of occupational health, 46(3), 175-180 (2004-06-25)
A method for the qualitative analysis of aromatic hydrocarbons in air and their various urinary metabolites is presented. The air was sampled in charcoal tubes and extracted with carbon disulfide. The hydrocarbons were identified as being aliphatic hydrocarbons (C(9)-C(19)), aromatic
S Fujisawa et al.
Journal of dental research, 82(1), 43-47 (2003-01-01)
The liquid of 2-ethoxybenzoic acid cements is composed of 2-ethoxybenzoic acid and eugenol (4-allyl-2-methoxyphenol). Recently, eugenol was reported to produce radicals at a higher pH, which consequently directly damages cells. We examined here whether eugenol radicals are generated from the
Chun-Pin Lin et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 71(2), 429-440 (2004-09-25)
Various root-end filling materials have been used to prevent the entry of root-canal pathogens into periapical regions. Five root-end filling materials were compared regarding the cytotoxicity, apoptosis, and mitochondrial dehydrogenase (MDH) activities of human periodontal ligament (PDL) fibroblasts, with the
A Timmerman et al.
Australian dental journal, 62(3), 345-354 (2017-03-09)
This study describes the cross sectional and longitudinal data of endodontic and periapical status of new patients presenting to a major dental hospital, and assesses the relationships between tooth-related variables with apical periodontitis. The records of 695 patients were randomly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service