Skip to Content
Merck
All Photos(2)

Key Documents

146668

Sigma-Aldrich

2,4-Dichlorobenzyl alcohol

99%

Synonym(s):

(2,4-Dichlorophenyl)methanol, 2,4-Dichlorobenzenemethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3CH2OH
CAS Number:
Molecular Weight:
177.03
Beilstein:
1448652
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

mp

55-58 °C (lit.)

functional group

chloro
hydroxyl

SMILES string

OCc1ccc(Cl)cc1Cl

InChI

1S/C7H6Cl2O/c8-6-2-1-5(4-10)7(9)3-6/h1-3,10H,4H2

InChI key

DBHODFSFBXJZNY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4-Dichlorobenzyl alcohol is a constituent of commercially available lozenges for acute sore throat caused by upper respiratory tract infections.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alex Thompson et al.
Drugs in R&D, 13(2), 101-107 (2013-04-17)
Acute sore throat (pharyngitis) is one of the most common illnesses for which children are seen by primary care physicians. Most cases are caused by viruses and are benign and self-limiting. Clinically proven, over-the-counter throat lozenges provide rapid and effective

Articles

This application note describes the LC-MS-based quantitative analysis of known nitrosamine impurities following procedures 1 and 3 as given in USP general chapter <1469>.

This application note describes the LC-MS-based quantitative analysis of known nitrosamine impurities following procedures 1 and 3 as given in USP general chapter <1469>.

This application note describes the LC-MS-based quantitative analysis of known nitrosamine impurities following procedures 1 and 3 as given in USP general chapter <1469>.

This application note describes the LC-MS-based quantitative analysis of known nitrosamine impurities following procedures 1 and 3 as given in USP general chapter <1469>.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service