145424
5-Acetoxymethyl-2-furaldehyde
99%
Synonym(s):
(5-Formylfuran-2-yl)methyl acetate, 5-(Acetoxymethyl)-2-furaldehyde, 5-(Acetoxymethyl)-2-furancarboxaldehyde, 5-(Acetoxymethyl)furfural, 5-(Acetoxymethyl)furfuraldehyde
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H8O4
CAS Number:
Molecular Weight:
168.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
form
solid
mp
53-55 °C (lit.)
SMILES string
[H]C(=O)c1ccc(COC(C)=O)o1
InChI
1S/C8H8O4/c1-6(10)11-5-8-3-2-7(4-9)12-8/h2-4H,5H2,1H3
InChI key
QAVITTVTXPZTSE-UHFFFAOYSA-N
General description
5-Acetoxymethyl-2-furaldehyde is a volatile flavor compound present in berrycactus. It is a potential sweet modulator compound present in traditional balsamic vinegar of Modena.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Moises Alejandro Vazquez-Cruz et al.
Journal of food science, 77(4), C366-C373 (2012-03-21)
Descriptive sensory analysis combined with a chemical analysis, provided insight regarding sensory significance for a better understanding of berrycactus' flavor. This study was based on the volatile components derived from the gas chromatography-olfactometry (GC-O) analysis. Four different ripening stages were
Hedda Hillmann et al.
Journal of agricultural and food chemistry, 60(40), 9974-9990 (2012-09-14)
Sensory-directed fractionation of traditional balsamic vinegar of Modena (TBV) led to the identification of the sweet-bitter tasting hexose acetates 6-O-acetyl-α/β-d-glucopyranose and 1-O-acetyl-β-d-fructopyranose as well as the previously unknown sweetness modulator 5-acetoxymethyl-2-furaldehyde. Taste re-engineering experiments and sensory time-intensity studies confirmed 5-acetoxymethyl-2-furaldehyde
Pak Hin Chow et al.
Molecular pharmacology, 98(1), 38-48 (2020-05-22)
Aquaporin-1 (AQP1) dual water and ion channels enhance migration and invasion when upregulated in leading edges of certain classes of cancer cells. Work here identifies structurally related furan compounds as novel inhibitors of AQP1 ion channels. 5-Hydroxymethyl-2-furfural (5HMF), a component
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service