Skip to Content
Merck
All Photos(1)

Key Documents

14527

Sigma-Aldrich

O,O′-Bis(2-aminopropyl) polypropylene glycol-block-polyethylene glycol-block-polypropylene glycol

Mr 800

Synonym(s):

Jeffamine® ED-900, Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(NH2)CH2[OCH(CH3)CH2]l(OCH2CH2)m[OCH2CH(CH3)]nNH2
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

product name

O,O′-Bis(2-aminopropyl) polypropylene glycol-block-polyethylene glycol-block-polypropylene glycol1, 800

Quality Level

mol wt

Mr ~900

General description

Jeffamine® ED-900 is an amine-terminated poly(ethylene glycol)/poly(propylene glycol) copolymer. It exists as a low viscosity and low vapor pressure liquid.{4}

Application

Jeffamine® ED-900 may be used to fabricate thin electrolyte films in diureasil matrix.{5} Succinyl chloride polymerized with ED-900 may be used as a drug carrier.{6}

Legal Information

Jeffamine is a registered trademark of Huntsman Petrochemical, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of copolymers by laser desorption fourier transform mass spectrometry.
Nuwaysir L, et al.
Journal of the American Society For Mass Spectrometry, 1 (1), 66-71 (1990)
Water?soluble polyamides as potential drug carriers, VII. Synthesis of polymers containing intrachain?or extrachain?type amine ligands by interfacial polymerization.
Chiba U, et al.
Macromolecular Materials and Engineering, 214(1), 137-152 (1994)
Nuwaysir LM, et al.
Journal of the American Chemical Society null
Studies of solid-state electrochromic devices based on PEO/siliceous hybrids doped with lithium perchlorate.
Barbosa PC, et al.
Electrochimica Acta, 52(8), 2938-2943 (2007)
Jun Pan et al.
Journal of biomedical materials research. Part A, 89(1), 160-167 (2008-04-24)
A new poly(ethylene glycol) (PEG)-modified poly(D,L-lactic acid) (PLA) was synthesized by grafting maleic anhydride onto PLA and subsequently amidating with O,O'-bis-(2-aminopropyl) polypropylene glycol-block-polyethylene oxide-block-polypropylene glycol (H2N-PEG-NH2, Mw: 600). Its structure was confirmed by FTIR, DSC, 1H NMR, GPC, and ninhydrin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service