Skip to Content
Merck
All Photos(2)

Key Documents

144800

Sigma-Aldrich

Tetrabutylphosphonium chloride

greener alternative

96%

Synonym(s):

Tetra-n-butylphosphonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4P(Cl)
CAS Number:
Molecular Weight:
294.88
Beilstein:
4019186
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

liquid

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

62-66 °C (dec.) (lit.)

greener alternative category

SMILES string

[Cl-].CCCC[P+](CCCC)(CCCC)CCCC

InChI

1S/C16H36P.ClH/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;/h5-16H2,1-4H3;1H/q+1;/p-1

InChI key

IBWGNZVCJVLSHB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application


  • Insight into Cellulose Dissolution with the Tetrabutylphosphonium Chloride-Water Mixture using Molecular Dynamics Simulations.: This study investigates the molecular dynamics of cellulose dissolution in a Tetrabutylphosphonium chloride-water mixture, providing insights into its potential application in biomass processing and material science (Crawford and Ismail, 2020).

  • Thermodynamic and Transport Properties of Tetrabutylphosphonium Hydroxide and Tetrabutylphosphonium Chloride-Water Mixtures via Molecular Dynamics Simulation.: The research focuses on the thermodynamic and transport properties of Tetrabutylphosphonium chloride in water mixtures, highlighting its relevance in developing advanced ionic liquids for various industrial applications (Crawford and Ismail, 2020).

  • End-of-life Treatment of Poly(vinyl chloride) and Chlorinated Polyethylene by Dehydrochlorination in Ionic Liquids.: This article explores the use of Tetrabutylphosphonium chloride as an ionic liquid for the dehydrochlorination of PVC and CPE, demonstrating its utility in sustainable polymer recycling processes (Glas et al., 2014).

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1C - Skin Sens. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

>266.0 °F - closed cup

Flash Point(C)

> 130 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenbao Ma et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(30), 7287-7296 (2017-04-06)
The efficient and environmentally benign epoxidation of allylic alcohols has been attained by using new kinds of monomeric peroxotantalate anion-functionalized ionic liquids (ILs=[P
Mitsuru Abe et al.
Chemical communications (Cambridge, England), 48(12), 1808-1810 (2012-01-06)
Tetrabutylphosphonium hydroxide containing 40 wt% water dissolved 20 wt% cellulose at the final concentration within 5 minutes under mild stirring at 25 °C.
Shohei Saita et al.
Chemical communications (Cambridge, England), 49(1), 93-95 (2012-11-21)
The phase separation temperature of the tetrabutylphosphonium trifluoroacetate-water mixture, which undergoes a lower critical solution temperature-type phase transition, is highly sensitive to the hydrophilicity of added salts; this system can be used to compare the hydrophilicity of the added ions.
Kenta Fukumoto et al.
Chemical communications (Cambridge, England), (29)(29), 3081-3083 (2006-07-21)
Hydrophobic ionic liquids composed of tetrabutylphosphonium cation and chiral anions derived from amino acids modified with trifluoromethane sulfonyl groups have been synthesized using a simple method.
Junko Kagimoto et al.
Chemical communications (Cambridge, England), (21)(21), 2254-2256 (2006-05-24)
Tetraalkylphosphonium-based amino-acid ionic liquids show lower viscosities and higher decomposition temperatures (>300 degrees C) than previously reported ammonium-based amino-acid ionic liquids.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service