Skip to Content
Merck
All Photos(1)

Key Documents

142905

Sigma-Aldrich

Lapachol

98%

Synonym(s):

2-Hydroxy-3-(3-methyl-2-butenyl)-1,4-naphthoquinone, Natural Yellow 16

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14O3
CAS Number:
Molecular Weight:
242.27
Colour Index Number:
75490
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

141-143 °C (lit.)

solubility

ethanol: soluble 10 mg/mL, clear, light yellow to yellow

functional group

ketone

SMILES string

C\C(C)=C\CC1=C(O)C(=O)c2ccccc2C1=O

InChI

1S/C15H14O3/c1-9(2)7-8-12-13(16)10-5-3-4-6-11(10)14(17)15(12)18/h3-7,18H,8H2,1-2H3

InChI key

CIEYTVIYYGTCCI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lapachol is natural naphthoquinone compound derived from Bignoniaceae (Tabebuia sp.).

Application

Lapachol was used in the synthesis of the lapachol metal complexes.

Biochem/physiol Actions

Lapachol has antimicrobial properties against many pathogens. It has anti-inflammatory, analgesic and antibiotic properties. It is inhibitor of epithelial tumors in Drosophila melanogaster heterozygote.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xueyin Zu et al.
Phytotherapy research : PTR, 33(9), 2337-2346 (2019-06-22)
Lapachol is a 1,4-naphthoquinone that is isolated from the Bignoniaceae family. It has been reported to exert anti-inflammatory, antibacterial, and anticancer activities. However, the anticancer activity of lapachol and its molecular mechanisms against esophageal squamous cell carcinoma (ESCC) cells have
Ingrid L Cockburn et al.
Biological chemistry, 392(5), 431-438 (2011-03-24)
Plasmodium falciparum heat shock protein 70 (PfHsp70-1) is thought to play an essential role in parasite survival and virulence in the human host, making it a potential antimalarial drug target. A malate dehydrogenase based aggregation suppression assay was adapted for
Kenneth O Eyong et al.
Bioorganic & medicinal chemistry letters, 18(20), 5387-5390 (2008-10-03)
Ozonolysis of lapachol (1), resulting in an unusual formation of a potent antitumor agent 2-acetylfuranonaphthoquinone (3) along with the expected aldehyde 6, is described. The reaction of lapachol (1) with CAN in dry acetonitrile leading to biologically active furanonaphthoquinones is
Camila Raquel Paludo et al.
European journal of drug metabolism and pharmacokinetics, 42(2), 213-220 (2016-04-10)
β-Lapachone is a drug candidate in phase II clinical trials for treatment of solid tumors. The therapeutic efficacy of β-lapachone is closely related to its metabolism, since this o-naphthoquinone produces cytotoxic effect after intracellular bioreduction by reactive oxygen species formation.
Ricardo Vessecchi et al.
Rapid communications in mass spectrometry : RCM, 24(14), 2101-2108 (2010-06-17)
Electrospray ionization mass spectrometric analysis of lapachol (2-hydroxy-3-(3-methyl-2-butenyl)-1,4-naphthoquinone) was accomplished in order to elucidate the gas-phase dissociation reactions of this important biologically active natural product. The occurrence of protonated and cationized species in the positive mode and of deprotonated species

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service