Skip to Content
Merck
All Photos(1)

Key Documents

141038

Sigma-Aldrich

2-Hydroxybenzonitrile

99%

Synonym(s):

2-Cyanophenol, Salicylonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCC6H4OH
CAS Number:
Molecular Weight:
119.12
Beilstein:
1210029
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

bp

149 °C/14 mmHg (lit.)

mp

92-95 °C (lit.)

functional group

nitrile

SMILES string

Oc1ccccc1C#N

InChI

1S/C7H5NO/c8-5-6-3-1-2-4-7(6)9/h1-4,9H

InChI key

CHZCERSEMVWNHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Hydroxybenzonitrile may be used in the preparation of:
  • methyl-(2-cyanophenoxy)acetate
  • 3-amino-N-phenylbenzofuran-2-carboxamide
  • 2-(2-oxo-2-phenylethoxy)benzonitrile
  • 3-amino-2-cyanobenzo[b]furane
It may also be used as a directing group for meta-selective C-H bond alkenylation.
2-Hydroxybenzonitrile was used as starting reagent during the synthesis of:
  • mono-alkoxyphenyloxazoline
  • furanoside

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective Cu-catalyzed 1, 4-addition of Me3Al to a 4, 4-disubstituted cyclohexa-2, 5-dienone.
Takemoto Y, et al.
Tetrahedron, 52(45), 14177-14188 (1996)
Reactivity of 1, 2-cyclic sulfite xylosides towards nucleophiles.
Batoux N, et al.
Tetrahedron, 65(43), 8858-8862 (2009)
Efficient New Synthesis of N-Arylbenzo [b] furo [3, 2-d] pyrimidin-4-amines and Their Benzo [b] thieno [3, 2-d] pyrimidin-4-amine Analogues via a Microwave-Assisted Dimroth Rearrangement.
Loidreau Y, et al.
Journal of Heterocyclic Chemistry, 50(5), 1187-1197 (2013)
meta-Selective arene C-H bond olefination of arylacetic acid using a nitrile-based directing group.
Bera M, et al.
Organic Letters, 16(21), 5760-5763 (2014)
A microwave-assisted multicomponent protocol for the synthesis of benzofuran-2-carboxamides.
Vincetti P, et al.
Tetrahedron Letters, 57(13), 1464-1467 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service