Skip to Content
Merck
All Photos(1)

Key Documents

139300

Sigma-Aldrich

Ethyl 4-nitrocinnamate, predominantly trans

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4CH=CHCO2C2H5
CAS Number:
Molecular Weight:
221.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

mp

138-140 °C (lit.)

functional group

ester
nitro

SMILES string

CCOC(=O)\C=C\c1ccc(cc1)[N+]([O-])=O

InChI

1S/C11H11NO4/c1-2-16-11(13)8-5-9-3-6-10(7-4-9)12(14)15/h3-8H,2H2,1H3/b8-5+

InChI key

PFBQVGXIMLXCQB-VMPITWQZSA-N

Related Categories

Application

Ethyl 4-nitrocinnamate was used to study the kinetics of reduction of ethyl t-cinnamate and its substituted derivatives by samarium diiodide in the presence of hexamethylphosphoramide and t-butanol.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reduction of Ethyl t-cinnamates by Samarium Diiodide
Lin T-Y, et al.
J. Chin. Chem. Soc., 48(5) (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service