Skip to Content
Merck
All Photos(1)

Key Documents

13454

Sigma-Aldrich

N-Benzylhydroxylamine hydrochloride

puriss., ≥99.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2NHOH · HCl
CAS Number:
Molecular Weight:
159.61
Beilstein:
507948
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

puriss.

Quality Level

Assay

≥99.0% (AT)

form

solid

mp

~105 °C
108-110 °C (lit.)

functional group

amine
phenyl

SMILES string

Cl.ONCc1ccccc1

InChI

1S/C7H9NO.ClH/c9-8-6-7-4-2-1-3-5-7;/h1-5,8-9H,6H2;1H

InChI key

YSNXOQGDHGUKCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

N-Benzylhydroxylamine hydrochloride was used in the synthesis of sugar derived nitrones. It was used as starting reagent in the synthesis of fluoro isoxazoline and isoxazolidine derivatives using flouro nitrone.

Biochem/physiol Actions

N-Benzylhydroxylamine is a potential pharmacological agent in the prevention and progression of acrolein-induced damage to the retinal pigment epithelium.

Other Notes

Formation of nitrones from carbonyl compounds and [2+3]-cycloaddition with olefins to isoxazolidines, which are versatile intermediates; Pd-catalyzed hydroxylamination of allylic esters.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B.J. Wakefield
Sci. Synth., 11, 229-229 (2002)
T. Kawakami et al.
Bulletin of the Chemical Society of Japan, 73, 2423-2423 (2000)
R Herrera et al.
The Journal of organic chemistry, 66(4), 1252-1263 (2001-04-21)
Captodative olefins 1-acetylvinyl carboxylates proved to be highly regioselective dipolarophiles in 1,3-dipolar cycloadditon to propionitrile oxide, arylphenylnitrile imines, diazoalkanes, and nitrones to yield the corresponding 5-substituted heterocycles. The addition of the latter was also stereoselective, being slightly susceptible to steric
Ionic liquid mediated synthesis of some novel fluoro isoxazolidine and isoxazoline derivatives using N-benzyl fluoro nitrone via cycloaddition reaction and their antimicrobial activities.
Chakraborty B, et al.
Indian J. Chem. B, 52(10), 1342-1351 (2013)
J.E. Baldwin et al.
Tetrahedron, 42, 3097-3097 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service