134198
5-Methyl-3-phenylisoxazole-4-carboxylic acid
99%
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Assay
99%
mp
192-194 °C (lit.)
functional group
carboxylic acid
phenyl
SMILES string
Cc1onc(-c2ccccc2)c1C(O)=O
InChI
1S/C11H9NO3/c1-7-9(11(13)14)10(12-15-7)8-5-3-2-4-6-8/h2-6H,1H3,(H,13,14)
InChI key
PENHKTNQUJMHIR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
5-Methyl-3-phenylisoxazole-4-carboxylic acid was used in preparation of intermediates for the synthesis of penicillin. It was used for acylation during solid support synthesis of the isoxazolopyridone derivatives.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Bioorganic & medicinal chemistry letters, 20(2), 726-729 (2009-12-17)
This Letter describes the synthesis and evaluation of mGluR7 antagonists in the isoxazolopyridone series. In the course of modification in this class, novel solid support synthesis of the isoxazolopyridone scaffold was developed. Subsequent chemical modification led to the identification of
1112. Derivatives of 6-aminopenicillanic acid. Part VI. Penicillins from 3-and 5-phenylisoxazole-4-carboxylic acids and their alkyl and halogen derivatives.
Journal of the Chemical Society, 5838-5845 (1963)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service