Skip to Content
Merck
All Photos(2)

Key Documents

134163

Sigma-Aldrich

1-(4-Sulfophenyl)-3-methyl-5-pyrazolone

Synonym(s):

3-Methyl-1-(4-sulfophenyl)-2-pyrazolin-5-one, 4-(3-Methyl-5-oxo-2-pyrazolin-1-yl)benzenesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10N2O4S
CAS Number:
Molecular Weight:
254.26
Beilstein:
619424
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

mp

>252 °C (dec.) (lit.)

functional group

sulfonic acid

SMILES string

CC1=NN(C(=O)C1)c2ccc(cc2)S(O)(=O)=O

InChI

1S/C10H10N2O4S/c1-7-6-10(13)12(11-7)8-2-4-9(5-3-8)17(14,15)16/h2-5H,6H2,1H3,(H,14,15,16)

InChI key

CWJQQASJVVAXKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-(4-Sulfophenyl)-3-methyl-5-pyrazolone (4-(3-methyl-5-oxo-2-pyrazolin-1-yl) benzoic acid) was used for pre-column derivatization of carbohydrates. It was also used as derivatization reagent in determination of oligosides, mannose and galactose obtained by degradation of galactomannans.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Precolumn derivatization of reducing carbohydrates with 4-(3-methyl-5-oxo-2-pyrazolin-1-yl) benzoic acid. Study of reaction, high-performance liquid chromatographic separation and quantitative performance of method.
Castells CB, et al.
Chromatographia, 56(3-4), 153-160 (2002)
N Tapie et al.
Journal of chromatography. A, 1181(1-2), 45-50 (2008-01-08)
The present work describes the validation of an easy, fast and efficient precolumn derivatization method for the quantification of oligosides, mannose and galactose obtained by degradation of galactomannans. This work combines an acid hydrolysis and an enzymatic degradation of natural

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service