Skip to Content
Merck
All Photos(1)

Key Documents

13282

Sigma-Aldrich

Benzyl chloromethyl ether

technical, ~60% (NMR)

Synonym(s):

(Chloromethoxymethyl)benzene, Benzyloxymethyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OCH2Cl
CAS Number:
Molecular Weight:
156.61
Beilstein:
1364034
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Quality Level

Assay

~60% (NMR)

form

liquid

impurities

~10% formaldehyde dibenzylacetal
~25% benzyl chloride

refractive index

n20/D 1.527

bp

102 °C/14 mmHg (lit.)

density

1.126 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

ClCOCc1ccccc1

InChI

1S/C8H9ClO/c9-7-10-6-8-4-2-1-3-5-8/h1-5H,6-7H2

InChI key

LADPCMZCENPFGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Acid protecting group; Introduction of benzoyloxy- and hydroxymethyl groups

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral - STOT SE 3

Target Organs

Heart,forestomach, Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D.S. Connor et al.
Organic Syntheses, 52, 16-16 (1972)
P.A. Zoretic et al.
The Journal of Organic Chemistry, 40, 2962-2962 (1975)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service