Skip to Content
Merck
All Photos(2)

Key Documents

131849

Sigma-Aldrich

1,2,3,4-Tetrachlorobenzene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H2Cl4
CAS Number:
Molecular Weight:
215.89
Beilstein:
1910025
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

chunks

bp

254 °C/761 mmHg (lit.)

mp

42-47 °C (lit.)

functional group

chloro

SMILES string

Clc1ccc(Cl)c(Cl)c1Cl

InChI

1S/C6H2Cl4/c7-3-1-2-4(8)6(10)5(3)9/h1-2H

InChI key

GBDZXPJXOMHESU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,2,3,4-tetrachlorobenzene was used as a model compound to determine the chlorobenzenes (CBs) in soil samples. It was also used for rapid dechlorination of 1,2,3,4-tetrachlorodibenzo-p-dioxin (1,2,3,4-TeCDD).

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Young-Beom Ahn et al.
FEMS microbiology ecology, 66(2), 271-281 (2008-07-22)
Dechlorination of spiked 1,2,3,4-tetrachlorodibenzo-p-dioxin (1,2,3,4-TeCDD) was investigated in sediment microcosms from three polychlorinated dibenzo-p-dioxin and dibenzofuran (CDD/F)-contaminated sites: River Kymijoki, Finland; Gulf Island Pond, Maine; and Lake Roosevelt, Washington. Dechlorination was stimulated by addition of electron donor and halogenated priming
Li Wang et al.
Journal of chromatography. A, 1256, 9-14 (2012-08-15)
A novel and simple analytical method for the determination of chlorobenzenes (CBs) in soil samples was developed using ultrasonic-assisted water extraction (UAWE) coupled with solvent bar microextraction (SBME). Four chlorobenzenes, 1,2,3-trichlorobenzene (1,2,3-TCB), 1,2,3,4-tetrachlorobenzene (1,2,3,4-TeCB), hexachlorobenzene (HCB), and 1-chloro-4-nitrobenzene (1-C-4-NB), were
Ken G Drouillard et al.
Bulletin of environmental contamination and toxicology, 93(5), 561-566 (2014-06-18)
A vial equilibration technique was used to estimate the fugacity capacities of food and feces samples for 1,2,3,4-tetrachlorobenzene (TCB). The method was calibrated using different volumes of n-octanol and by comparing the measured and predicted fugacity capacity (Zoct) of n-octanol
B A Kimball et al.
Journal of wildlife diseases, 32(3), 505-511 (1996-07-01)
Pentachlorobenzene (PeCB) and 1,2,3,4-tetrachlorobenzene (TeCB) were evaluated as oral chemical biomarkers when administered to coyotes (Canis latrans) during the period of 31 January to 10 August 1994. Three coyotes each received 100 mg of PeCB and three received 100 mg
L D Bowers et al.
Clinical chemistry, 43(6 Pt 1), 1033-1039 (1997-06-01)
Limits of detection are important issues for GC/MS screening for anabolic agents and for confirmation of various drugs of abuse. We compared a quadrupole ion trap (QIT) operated in two different selected-ion storage modes and a quadrupole mass filter (QMF)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service